Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU158121

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Dtx3l

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTAACTCTGTTGACTCAGTTGTCCAAAAGATCTTTCTTGCTGTGACCGCTGAGCTGAACTGTGACCTGCTCTCTAAAGAGCAGAGAGCATCTATAACCACTGTCTGCCCTCACATCATCAAAAGCATGGAGGGTAGTGATGGAATTAAGAAGGTGTGTGGCAACTTCAAAGATATTGAAAAGATACATCACTTCTTGAGTGAGCAGCTTTTGGAACGGGAGCAGAAACGGAAGGGAAGCGAGCAGAAACGGAAGTGCGCCCCACAGAAACACACACCTCCCGATGTGGAGCGGGAGCCCCCTGATCAGAGCAGTATTCAAGTTCCTGTGCTTCTCCTTGAATATTTCAAGCATGTTAATCCGGGTAGACTAGAGTTCATAGAGTACAAATTTGGTGTAAACATTGAAATCCAAGCTAGTTCTCCCAATATGGTCACTGTAGGCTTCACCTCCAGCCCATTTGGCAACGTAGAAGAAGCAAGTCAGTCCTTTGTCAGAGACTTTCAGAAAT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danielle P Johnson et al.
Oncotarget, 6(7), 4863-4887 (2015-01-22)
Gain-of-function mutations in the catalytic site of EZH2 (Enhancer of Zeste Homologue 2), is observed in about 22% of diffuse large B-cell lymphoma (DLBCL) cases. Here we show that selective inhibition of histone deacetylase 1,2 (HDAC1,2) activity using a small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service