Skip to Content
MilliporeSigma
All Photos(1)

Documents

EMU152291

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Anks1b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGGCTGCTTTCTTTTCTTACCTGGAGATTGCACTGTGGTGGTGGCAAGGGAGAAATTAATGCCCACATGTTGAACTTAAGTCTTTTAAAGTTTAGCCCAGGTATCCCTACACTCCTTGTTCGAGTGACAAATCATTATAATATTCTTTCAAACAACTGTCATACCGAAATACCAAAGGCTATATTAGGTAAAACAGCAGACAGTATACATAGGATCAAGTAACCCTATTTATTTGTAATAGAATCCACCAAGGGGAAAGGATGCAAAATAAACTTAGAAGCATGAATGAAGTTAGGTGTGGGGTAGTCTGAATATCAGGCTAAGACTTCTGGTATTTATTCCTTAGGAAATGAGAAAAGCACAAAGGATTTGGGGCAGTTAAATGACATCATTGGAACTGGCCTTTCAGAGAATGAATGTGATAGAAACTGGTCATTGCATTTAGAAAACGTAATCCAATTTCATTGACCTTCTGTATGGCCTGCATCTTATTTGTATTCATCCTGTCT

shipped in

ambient

storage temp.

−20°C

Gene Information

mouse ... ANKS1B(77531)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S-C Tien et al.
Oncogene, 33(22), 2938-2946 (2013-06-19)
Deregulation of Shp2, a non-receptor tyrosine phosphatase, causes hyperactivation of extracellular signal-regulated kinase (ERK), leading to growth abnormality. Here, we show that inhibition of RhoA-Dia is sufficient to upregulate ERK activation in epithelial cells. Oncogenic Shp2 expression attenuates RhoA-Dia signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service