Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU148751

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Rhoa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTCATGCGGTTAATTTGAAGTGCTGTTTATTAATCTTAGTGTATGATTACTGGCCTTTTCATTTATCTATAATTTACCTAAGATTACAAATCAGAAGTCATCTTGCTACCAGTATTTAGAAGCCAACCACGATTATTAATAATGTCCAACCTGTCTGACCAGCCAGGGTCCTTCTGACACTGCTCTAACAGCCCTCTCTGCACTCCACCTGACACCAGGCGCTAATTCAAAGAATTTCTTAACTTCTTGCTTCTTTCTAGAAAGAGAAACAGTTGGTAACTTTTGTGAATTAGGCTGTAACTACTTTATAACTAACATGTCCTGCCTACTTTCTGTCAACTGCAAGAACTCTGGTGAGTCACTACTTCAGAGCTTTCCTTGTTAACAGACTCCATTGCCAGAGCTCTGGGGTGGGTATTCAGTTTTTTGAAATCTTGCTCAGCCAGAAAGGCCCAAGTCCACGCAGCTGTTGCAGAGTTACAGTTCTGTGGTCTCATGTTAGTTACCTT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurent Pieuchot et al.
Nature communications, 9(1), 3995-3995 (2018-09-30)
Cells have evolved multiple mechanisms to apprehend and adapt finely to their environment. Here we report a new cellular ability, which we term "curvotaxis" that enables the cells to respond to cell-scale curvature variations, a ubiquitous trait of cellular biotopes.
Anat Biran et al.
Genes & development, 29(8), 791-802 (2015-04-10)
Mammalian cells mostly rely on extracellular molecules to transfer signals to other cells. However, in stress conditions, more robust mechanisms might be necessary to facilitate cell-cell communications. Cellular senescence, a stress response associated with permanent exit from the cell cycle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service