Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU076591

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Atp7a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAAAATGCATCCCTGGTTCAAATTGATGCAATTAATGAACAGTCATCAACTTCATCTTCTATGATCATTGATGCTCATCTCTCAAATGCTGTTAATACTCAGCAGTACAAAGTCCTCATTGGTAACCGGGAATGGATGATTAGAAATGGTCTTGTCATAAGTAATGATGTAGATGAATCTATGATTGAACATGAAAGAAGAGGTCGGACTGCTGTCTTGGTGACAATCGATGATGAGCTGTGTGGCTTGATGGCTATTGCTGATACTGTGAAACCTGAGGCCGAGTTGGCTGTACACATTCTGAAATCTATGGGTTTAGAAGTAGTTCTGATGACTGGAGACAACAGTAAAACTGCTCAGTCTATTGCTTCTCAGGTTGGCATTACTAAGGTGTTTGCTGAAGTTCTACCTTCCCACAAAGTTGCTAAGGTGAAGCAGCTCCAAGA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huilian Huang et al.
Clinical laboratory, 61(10), 1501-1508 (2015-12-09)
Mounting evidence indicates that nuclear targeting by growth factors plays an indispensable role on their biological activities. Midkine (MK) is a multifunctional growth factor and has been discovered to play important roles in carcinogenesis. MK has been reported to localize
Xue Fu et al.
Toxicological sciences : an official journal of the Society of Toxicology, 139(2), 432-451 (2014-03-13)
Regulation of cellular copper (Cu) homeostasis involves Cu-transporting ATPases (Cu-ATPases), i.e., ATP7A and ATP7B. The question as to how these Cu-ATPases in brain barrier systems transport Cu, i.e., toward brain parenchyma, cerebrospinal fluid (CSF), or blood, remained unanswered. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service