Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU056071

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Mavs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TAGGAAGCCCTGAGCCACTAGCCACCCAGCAGCCCCAAGAAGAGGAAGAACATTGTGCCAGTTCAATGCCCTGGGCTAAGTGGCTTGGGGCCACCAGTGCACTCTTGGCTGTATTCCTGGCAGTGATGCTGTACCGTAGTAGGCGCCTGGCCCAGTGAAGCCTCAGCTGTATGCTGTTCTCTTGCTCAGTTCTGCCAAGCATGTTCTCTAGGCTTGGGCTAGTAGAGGCTGAGTCAGAGAAACTTAAATATGGCGAGGTCCACTGAGCTATCCAGGTAGATAGCTACACCAAGACGTCATCACTGTTGGGTGGGGGAGAGACATTGTTTTATCCTGGTTCATATGTCATCTTCTGGTCTTCAGCTTTTGGAGGCACTGTGTTACCTCCATTGCTCCTGACCTGCCCACGTGGCAGTGTAAGAGTTCATGCTCTGTGCTCCTAAGGAGGTATCTCCACCAGCTTTATCCCTGTTGGCCCAAGCCTGAAGATGAGGAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pradip B Devhare et al.
PloS one, 8(5), e63793-e63793 (2013-05-15)
Hepatitis E virus (HEV) is a major cause of enterically transmitted acute hepatitis in developing nations and occurs in sporadic and epidemic forms. The disease may become severe with high mortality (20%) among pregnant women. Due to lack of efficient
Charlotte Odendall et al.
Nature immunology, 15(8), 717-726 (2014-06-24)
Type I interferon responses are considered the primary means by which viral infections are controlled in mammals. Despite this view, several pathogens activate antiviral responses in the absence of type I interferons. The mechanisms controlling type I interferon-independent responses are
Huachen Gan et al.
Scientific reports, 5, 17916-17916 (2015-12-09)
Influenza A virus (IAV) targets airway epithelial cells and exploits the host cell machinery to replicate, causing respiratory illness in annual epidemics and pandemics of variable severity. The high rate of antigenic drift (viral mutation) and the putative antigenic shift

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service