Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU055771

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Aurkb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCAGAAGTTGGCTGAGAACAAGAGTCAGGGCTCCACTGCCTCGCAAGGATCCCAGAACAAGCAGCCTTTCACTATTGACAACTTTGAGATTGGGCGTCCTTTGGGCAAAGGCAAATTTGGAAACGTGTACTTGGCTCGGGAGAAGAAGAGCCGTTTCATCGTGGCACTCAAGATCCTCTTCAAGTCTCAGATTGAGAAGGAGGGGGTAGAGCACCAGCTTCGCCGAGAGATCGAAATCCAGGCGCACCTGAAACATCCCAACATCCTTCAACTCTACAACTACTTCTACGACCAGCAGAGGATCTACTTAATCCTGGAATACGCCCCTCGCGGGGAACTCTACAAGGAACTGCAGAAGAGTCGGACCTTCGATGAGCAGCGGACTGCCACGATCATGGAGGAACTGTCAGATGCCCTGACCTACTGCCACAAGAAGAAGGTAATTCACAGAGACATAAAGCCGGAGAACCTGCTGTTAGGTCTGCAGGGAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Madejón et al.
Journal of hepatology, 63(2), 312-319 (2015-03-04)
Chronic hepatitis C is a leading cause of chronic liver disease, cirrhosis and hepatocellular carcinoma. DNA methylation and histone covalent modifications constitute crucial mechanisms of genomic instability in human disease, including liver fibrosis and hepatocellular carcinoma. The present work studies
Kazuharu Kai et al.
Molecular cancer therapeutics, 14(12), 2687-2699 (2015-10-08)
Currently, no targeted drug is available for triple-negative breast cancer (TNBC), an aggressive breast cancer that does not express estrogen receptor, progesterone receptor, or HER2. TNBC has high mitotic activity, and, because Aurora A and B mitotic kinases drive cell
Lijuan Zhu et al.
The Journal of biological chemistry, 290(45), 27053-27066 (2015-09-18)
Mitotic chromosome segregation is orchestrated by the dynamic interaction of spindle microtubules with the kinetochores. During chromosome alignment, kinetochore-bound microtubules undergo dynamic cycles between growth and shrinkage, leading to an oscillatory movement of chromosomes along the spindle axis. Although kinetochore
Aarthi Jayanthan et al.
PloS one, 9(7), e102741-e102741 (2014-07-23)
Leukemia is the most common pediatric malignancy, constituting more than 30% of all childhood cancers. Although cure rates have improved greatly, approximately one in five children relapse and poor survival rates post relapse remain a challenge. Given this, more effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service