Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU026991

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Mdm2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTTTGGAGTCCCGAGTTTCTCTGTGAAGGAGCACAGGAAAATATATGCAATGATCTACAGAAATTTAGTGGCTGTAAGTCAGCAAGACTCTGGCACATCGCTGAGTGAGAGCAGACGTCAGCCTGAAGGTGGGAGTGATCTGAAGGATCCTTTGCAAGCGCCACCAGAAGAGAAACCTTCATCTTCTGATTTAATTTCTAGACTGTCTACCTCATCTAGAAGGAGATCCATTAGTGAGACAGAAGAGAACACAGATGAGCTACCTGGGGAGCGGCACCGGAAGCGCCGCAGGTCCCTGTCCTTTGATCCGAGCCTGGGTCTGTGTGAGCTGAGGGAGATGTGCAGCGGCGGCAGCAGCAGCAGTAGCAGCAGCAGCAGCGAGTCCACAGAGACGCCCTCGCATCAGGATCTTGACGATGGCGTAAGTGAGCATTCTGGTGATTGCCTGGATCAGGAT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaneabbas Raza et al.
Molecular and cellular biochemistry, 410(1-2), 187-195 (2015-09-10)
Estrogen is synthesized from cholesterol and high cholesterol levels are suggested to be associated with increased risk of estrogen receptor(ER)-positive breast cancer. The cholesterol metabolite 27-hydroxycholesterol (27-OHC) was recently identified as a selective estrogen receptor modulator (SERM) and may therefore
Eva Slabáková et al.
Oncotarget, 6(34), 36156-36171 (2015-09-30)
Plasticity of cancer cells, manifested by transitions between epithelial and mesenchymal phenotypes, represents a challenging issue in the treatment of neoplasias. Both epithelial-mesenchymal transition (EMT) and mesenchymal-epithelial transition (MET) are implicated in the processes of metastasis formation and acquisition of
Jiang-Jiang Qin et al.
Oncotarget, 6(5), 2623-2640 (2015-03-05)
The MDM2 oncogene has been suggested as a molecular target for treating human cancers, including breast cancer. Most MDM2 inhibitors under development are targeting the MDM2-p53 binding, and have little or no effects on cancers without functional p53, such as
Seemana Bhattacharya et al.
The FEBS journal, 281(13), 3061-3078 (2014-05-16)
Tumor suppressor retinoblastoma-associated protein (Rb) is an important cell cycle regulator, arresting cells in early G1. It is commonly inactivated in cancers and its level is maintained during the cell cycle. Rb is regulated by various post-translational modifications such as
Hong Zhu et al.
Oncotarget, 6(5), 3254-3267 (2014-09-17)
Adriamycin, a widely used anthracycline antibiotic in multiple chemotherapy regimens, has been challenged by the cardiotoxicity leading to fatal congestive heart failure in the worst condition. The present study demonstrated that Dihydromyricetin, a natural product extracted from ampelopsis grossedentat, exerted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service