Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU015391

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Map2k5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGATGTGATAGGCCAGGTTCTGCCTGAAGCGACGACGACAGCGTTTGAATATGAAGATGAAGATGGTGATAGGATTACAGTAAGAAGCGATGAAGAGATGAAGGCAATGCTGTCCTACTATTATTCCACAGTAATGGAACAGCAAGTAAATGGCCAGCTAATAGAGCCGCTGCAGATATTTCCAAGAGCCTGCAAGCCTCCCGGGGAACGGAACATACATGGCCTGAAGGTGAATACACGGGCTGGGCCATCTCAACACACCAGCCCTGTGGTCTCAGATTCGCTTCCAAGCAATAGCTTGAAGAAGTCCTCAGCTGAACTGAGAAAGATACTGGCCAACGGCCAGATGAATGAACAAGACATACGGTATCGAGACACCCTTGGTCATGGCAACGGAGGCACAGTCTACAAAGCACATCATGTCCCA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigeru Amano et al.
PloS one, 10(4), e0125054-e0125054 (2015-04-18)
The MEK/ERK pathways are critical for controlling cell proliferation and differentiation. In this study, we show that the MEK5/ERK5 pathway participates in osteoclast differentiation. ERK5 was activated by M-CSF, which is one of the essential factors in osteoclast differentiation. Inhibition
Shoichi Kaneshiro et al.
Biochemical and biophysical research communications, 463(3), 241-247 (2015-05-23)
Extracellular signal-regulated kinase 5 (ERK5) is a member of the mitogen-activated protein kinase (MAPK) family and is activated by its upstream kinase, MAPK kinase 5 (MEK5), which is a member of the MEK family. Although the role of MEK5 has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service