Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU007831

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Birc2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$292.00
50 μG
$534.00

$292.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$292.00
50 μG
$534.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$292.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTGTGTTCATTCCGTGTGGTCATCTAGTAGTCTGCCAGGAATGTGCCCCTTCTCTAAGGAAGTGCCCCATCTGCAGGGGGACAATCAAGGGGACTGTGCGCACATTTCTCTCATGAGTGAAGAATGGTCTGAAAGTATTGTTGGACATCAGAAGCTGTCAGAACAAAGAATGAACTACTGATTTCAGCTCTTCAGCAGGACATTCTACTCTCTTTCAAGATTAGTAATCTTGCTTTATGAAGGGTAGCATTGTATATTTAAGCTTAGTCTGTTGCAAGGGAAGGTCTATGCTGTTGAGCTACAGGACTGTGTCTGTTCCAGAGCAGGAGTTGGGATGCTTGCTGTATGTCCTTCAGGACTTCTTGGATTTGGAATTTGTGAAAGCTTTGGATTCAGGTGATGTGGAGCTCAGA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Lappas
Placenta, 35(10), 831-838 (2014-08-13)
Independent of their role in apoptosis, cellular inhibitors of apoptosis (cIAP) 1 and 2, have emerged as regulators of inflammation. Obesity in pregnancy is characterised by maternal and placental inflammation. Thus, the aim of this study was to determine the
Hong Jin et al.
Oncology research, 22(3), 167-176 (2015-07-15)
Emerging evidence suggests a potential role of cellular inhibitor of apoptosis protein 1 (cIAP1) in the development of human ovarian cancer. However, its function in the progression of ovarian cancer has not been clearly determined. Our study aimed to investigate
E-W Lee et al.
Cell death and differentiation, 22(9), 1463-1476 (2015-01-24)
Given their crucial role in apoptosis suppression, inhibitor of apoptosis proteins (IAPs) have recently become attractive targets for cancer therapy. Here, we report that cellular IAP2 (cIAP2) is specifically stabilized in several cancer cell lines, leading to resistance to Smac

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service