Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU003641

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Atp6ap2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$292.00
50 μG
$534.00

$292.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$292.00
50 μG
$534.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$292.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTCTTTCTCTCCGAACTGCAAGTGCTACATGATATTTCCAGTTTGTTGTCTCGTCATAAGCATCTAGCCAAGGACCATTCACCCGACTTGTATTCATTGGAGCTGGCAGGTTTGGATGAACTTGGGAAGCGTTATGGGGAAGACTCTGAACAGTTCAGGGATGCTTCTAAGATCCTTGTTGATGCTCTCCAAAAGTTTGCAGATGACATGTACAGTCTCTATGGTGGGAACGCAGTGGTAGAGTTAGTGACTGTCAAATCATTCGACACATCCCTTGTGAGGAAGTCAAGGACCATCCTTGAGGCAAAACAAGAGAACACCCAAAGTCCTTATAACCTTGCATATAAGTATAATTTGGAGTATTCAGTGGTTTTCAACTTGGTACTGTGGATTATGATCGGCTTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph C K Leung et al.
Apoptosis : an international journal on programmed cell death, 20(7), 907-920 (2015-03-27)
Glomerulo-podocytic communication plays an important role in the podocytic injury in IgA nephropathy (IgAN). In this study, we examine the role of podocytic angiotensin II receptor subtype 1 (AT1R) and prorenin receptor (PRR) in podocytic apoptosis in IgAN. Polymeric IgA
Wendy W Batenburg et al.
PloS one, 9(6), e100954-e100954 (2014-06-27)
Dysfunction of renin-angiotensin system (RAS) contributes to the pathogenesis of diabetic retinopathy (DR). Prorenin, the precursor of renin is highly elevated in ocular fluid of diabetic patients with proliferative retinopathy. Prorenin may exert local effects in the eye by binding
Feng Y Liu et al.
Journal of the renin-angiotensin-aldosterone system : JRAAS, 15(2), 99-108 (2014-03-05)
Since the discovery of the (pro)renin receptor (PRR), it has been considered as a novel bioactive molecule of the renin-angiotensin system (RAS). The activation of PRR can elicit a series of angiotensin II (AngII)-independent effects. In this study, we investigated
Caixia Li et al.
American journal of physiology. Endocrinology and metabolism, 309(3), E302-E310 (2015-06-18)
High glucose reduces autophagy and enhances apoptosis of podocytes. Previously, we reported that high glucose induced podocyte injury through upregulation of the (pro)renin receptor (PRR). We hypothesized that increasing PRR reduces autophagy and increases apoptosis of mouse podocytes exposed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service