Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU158111

Sigma-Aldrich

MISSION® esiRNA

targeting human TIA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCCGCTCCAAAGAGTACATATGAGTCAAATACCAAACAGCTATCATATGATGAGGTTGTAAATCAGTCTAGTCCAAGCAACTGTACTGTATACTGTGGAGGTGTTACTTCTGGGCTAACAGAACAACTAATGCGTCAGACTTTTTCACCATTTGGACAAATAATGGAAATTCGAGTCTTTCCAGATAAAGGATATTCATTTGTTCGGTTCAATTCCCATGAAAGTGCAGCACATGCAATTGTTTCTGTTAATGGTACTACCATTGAAGGTCATGTTGTGAAATGCTATTGGGGCAAAGAAACTCTTGATATGATAAATCCCGTGCAACAGCAGAATCAAATTGGATATCCCCAACCTTATGGCCAGTGGGGCCAGTGGTATGGAAATGCACAACAAATTGGCCAGTA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuzo Abe et al.
Cell & bioscience, 11(1), 122-122 (2021-07-05)
Tumor protein D52 (TPD52) reportedly plays an important role in the proliferation and metastasis of various cancer cells, including oral squamous cell carcinoma (OSCC) cells, and is expressed strongly at the center of the tumor, where the microenvironment is hypoxic.
Xuefeng Yang et al.
Biochimie, 154, 119-126 (2018-08-26)
Gastric cancer (GC) is one of the most common malignancies as well as the third leading cause for cancer-related death. Molecular basis of GC are essential and critical for its therapeutic treatment, but still remain poorly understood. T-cell intracellular antigen-1
Jovan Nikolic et al.
PLoS pathogens, 12(10), e1005942-e1005942 (2016-10-18)
Stress granules (SGs) are membrane-less dynamic structures consisting of mRNA and protein aggregates that form rapidly in response to a wide range of environmental cellular stresses and viral infections. They act as storage sites for translationally silenced mRNAs under stress
Alice Pasini et al.
Biochimica et biophysica acta. Gene regulatory mechanisms, 1861(5), 463-472 (2018-03-21)
Cyclooxygenase-2 (COX-2), with its main antifibrotic metabolite PGE2, is regarded as an antifibrotic gene. Repressed COX-2 expression and deficient PGE2 have been shown to contribute to the activation of lung fibroblasts and excessive deposition of collagen in pulmonary fibrosis. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service