Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU153211

Sigma-Aldrich

MISSION® esiRNA

targeting human TUBB3, RP11-566K11.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATTCTGGTGGACCTGGAACCCGGAACCATGGACAGTGTCCGCTCAGGGGCCTTTGGACATCTCTTCAGGCCTGACAATTTCATCTTTGGTCAGAGTGGGGCCGGCAACAACTGGGCCAAGGGTCACTACACGGAGGGGGCGGAGCTGGTGGATTCGGTCCTGGATGTGGTGCGGAAGGAGTGTGAAAACTGCGACTGCCTGCAGGGCTTCCAGCTGACCCACTCGCTGGGGGGCGGCACGGGCTCCGGCATGGGCACGTTGCTCATCAGCAAGGTGCGTGAGGAGTATCCCGACCGCATCATGAACACCTTCAGCGTCGTGCCCTCACCCAAGGTGTCAGACACGGTGGTGGAGCCCTACAACGCCACGCTGTCCATCCACCAGCTGGTGGAGAACACGGATGAGACCTACTGCATCGACAACGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sidika Öztop et al.
Anticancer research, 39(2), 655-662 (2019-02-04)
The challenges of cololorectal cancer (CRC) management include prediction of outcome and drug response or chemoresistance. This study aimed at examining whether βIII-tubulin (TUBB3), present in various types of normal tissues and cancer, is a biomarker for the response of
Yohei Sekino et al.
Anticancer research, 40(4), 1943-1951 (2020-04-03)
Targeted receptor tyrosine kinase inhibitor (TKI) is a standard treatment in advanced renal cell carcinoma (RCC). However, the role of PTEN in TKI resistance remains poorly understood. We aimed to determine the functional role of PTEN knockout and analyse the
Yohei Sekino et al.
Oncology, 98(10), 689-698 (2020-06-26)
βIII-Tubulin, encoded by the TUBB3 gene, is a microtubule protein. Several studies have shown that overexpression of TUBB3 is linked to poor prognosis and is involved in taxane resistance in some cancers. The aim of this study was to analyze
Mihoko A Tame et al.
Oncotarget, 8(42), 71536-71547 (2017-10-27)
Microtubules are cellular targets for a variety of anticancer therapies because of their critical function in mitosis. Taxol belongs to a class of microtubule targeting agents that suppresses microtubule dynamics and interferes with the functioning of the mitotic spindle, thereby
Qian Liu et al.
Aging, 12(4), 3713-3729 (2020-02-29)
P-glycoprotein (P-gp) and βIII-tubulin overexpression-mediated drug resistance leads to clinical therapy failure for paclitaxel. However, the development of paclitaxel-resistance reversal agents has not had much success. In this study, EM-E-11-4, a lathyrane-type diterpenoid extracted from Euphorbia micractina, demonstrated good anti-MDR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service