Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU148011

Sigma-Aldrich

MISSION® esiRNA

targeting human KIFC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCACGCAGCCACAGTGTATTCCAGCTACAGATTTCTGGGGAGCACTCCAGCCGAGGCCTGCAGTGTGGGGCCCCCCTCAGTCTTGTGGACCTGGCCGGGAGTGAGCGACTTGACCCCGGCTTAGCCCTCGGCCCCGGGGAGCGGGAACGCCTTCGGGAAACACAGGCCATTAACAGCAGCCTGTCCACGCTGGGGCTGGTTATCATGGCCCTGAGCAACAAGGAGTCCCACGTGCCTTACCGGAACAGCAAACTGACCTACCTGCTGCAGAACTCTCTGGGTGGTAGTGCTAAGATGCTCATGTTTGTGAACATTTCTCCACTGGAAGAGAACGTCTCCGAGTCCCTCAACTCTCTACGCTTTGCCTCCAAGGTGAACCAGTGTGTTATTGGTACTGCTCAGGCCAACAGGAAGTGAAGACGGATCCAGATCTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTGTCCC

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerhard Jungwirth et al.
Cancers, 11(4) (2019-04-18)
Kinesins play an important role in many physiological functions including intracellular vesicle transport and mitosis. The emerging role of kinesins in different cancers led us to investigate the expression and functional role of kinesins in meningioma. Therefore, we re-analyzed our
V Pannu et al.
Cell death & disease, 5, e1538-e1538 (2014-11-21)
Classical anti-mitotic drugs have failed to translate their preclinical efficacy into clinical response in human trials. Their clinical failure has challenged the notion that tumor cells divide frequently at rates comparable to those of cancer cells in vitro and in
Takeharu Imai et al.
Pathology, research and practice, 213(11), 1388-1393 (2017-10-02)
Esophageal squamous cell carcinoma (ESCC) is one of the most common human cancers. We previously reported that KIFC1 is involved in gastric cancer pathogenesis and that KIFC1 plays an important role in gastric cancer spheroid colony formation. However, the significance
Xing Wang et al.
Oncology letters, 18(6), 5739-5746 (2019-12-04)
Hepatocellular carcinoma (HCC) is a common type of malignant tumor worldwide with a high mortality rate. In the past 20 years, the morbidity rate of HCC has increased. Progress has been made in the clinical diagnosis and therapy for HCC.
Xiaowei Fu et al.
International journal of oncology, 52(6), 1912-1922 (2018-04-06)
Kinesin family member C1 (KIFC1, also known as HSET) is a minus end-directed motor protein, which is critical in centrosome clustering. The present study investigated the expression of KIFC1 in paired hepatocellular carcinoma (HCC) tissues and adjacent non-cancerous tissues from 91 patients by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service