Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU143071

Sigma-Aldrich

MISSION® esiRNA

targeting human BAG3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGCAAAGAGGTGGATTCTAAACCTGTTTCCCAGAAGCCCCCACCTCCCTCTGAGAAGGTAGAGGTGAAAGTTCCCCCTGCTCCAGTTCCTTGTCCTCCTCCCAGCCCTGGCCCTTCTGCTGTCCCCTCTTCCCCCAAGAGTGTGGCTACAGAAGAGAGGGCAGCCCCCAGCACTGCCCCTGCAGAAGCTACACCTCCAAAACCAGGAGAAGCCGAGGCTCCCCCAAAACATCCAGGAGTGCTGAAAGTGGAAGCCATCCTGGAGAAGGTACAGGGGCTGGAGCAGGCTGTAGACAACTTTGAAGGCAAGAAGACTGACAAAAAGTACCTGATGATCGAAGAGTATTTGACCAAAGAGCTGCTGGCCCTGGATTCAGTGGACCCCGAGGGACGAGCCGATGTGCGTCAGGCCAGGAGAGACGGTGTCAGGAAGGTTCAGACCATCTTGGAAAAACTTGAACAGAAAGCCATTGATGTCCCAGGTCAAGTCCAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph M McClung et al.
Circulation, 136(3), 281-296 (2017-04-27)
Critical limb ischemia is a manifestation of peripheral artery disease that carries significant mortality and morbidity risk in humans, although its genetic determinants remain largely unknown. We previously discovered 2 overlapping quantitative trait loci in mice, We treated mice with
Patrick Antonietti et al.
Molecular cancer therapeutics, 16(1), 156-168 (2016-10-26)
Malignant gliomas exhibit a high intrinsic resistance against stimuli triggering apoptotic cell death. HSF1 acts as transcription factor upstream of HSP70 and the HSP70 co-chaperone BAG3 that is overexpressed in glioblastoma. To specifically target this resistance mechanism, we applied the
Fei Song et al.
Oncotarget, 8(56), 95392-95400 (2017-12-10)
Bcl2-associated athanogene 3 (BAG3) has been reported to be involved in aggressive progression of many tumors. In the present study, we examined the expression of BAG3 in human cervical cancer (CC) tissues and investigated the role of BAG3 in SiHa
Farzaneh G Tahrir et al.
Journal of cellular physiology, 232(4), 797-805 (2016-07-07)
Mitochondrial abnormalities impact the development of myofibrillar myopathies. Therefore, understanding the mechanisms underlying the removal of dysfunctional mitochondria from cells is of great importance toward understanding the molecular events involved in the genesis of cardiomyopathy. Earlier studies have ascribed a
Shuang Qiu et al.
Oncology reports, 38(1), 309-316 (2017-06-20)
Ovarian cancer is the most lethal disease among all gynecological malignancies. Interval cytoreductive surgery and cisplatin‑based chemotherapy are the recommended therapeutic strategies. However, acquired resistance to cisplatin remains a big challenge for the overall survival and prognosis in ovarian cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service