Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU142651

Sigma-Aldrich

MISSION® esiRNA

targeting human RSAD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGAGCAATGGAAGCCTGATCCGGGAGAGGTGGTTCCAGAATTATGGTGAGTATTTGGACATTCTCGCTATCTCCTGTGACAGCTTTGACGAGGAAGTCAATGTCCTTATTGGCCGTGGCCAAGGAAAGAAGAACCATGTGGAAAACCTTCAAAAGCTGAGGAGGTGGTGTAGGGATTATAGAGTCGCTTTCAAGATAAATTCTGTCATTAATCGTTTCAACGTGGAAGAGGACATGACGGAACAGATCAAAGCACTAAACCCTGTCCGCTGGAAAGTGTTCCAGTGCCTCTTAATTGAGGGTGAGAATTGTGGAGAAGATGCTCTAAGAGAAGCAGAAAGATTTGTTATTGGTGATGAAGAATTTGAAAGATTCTTGGAGCGCCACAAAGAAGTGTCCTGCTTGGTGCCTGAATCTAACCAGAAGATGAAAGACTCCTACCTTATTCTGGATGAATATATGCGCTTTCTGAACTGTAGAAAGGGACGGAAGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Wang et al.
Placenta, 36(6), 667-673 (2015-03-31)
Viperin, a virus-inducible antiviral protein, has been reported to inhibit the replication of a variety of viruses. However, its expression and function in trophoblast cells remains unclear. Toll-like receptor 3 (TLR3) is a key component of the innate immune system
Keaton M Crosse et al.
Immunology and cell biology, 99(4), 373-391 (2020-11-02)
Viperin is an interferon-inducible protein that is pivotal for eliciting an effective immune response against an array of diverse viral pathogens. Here we describe a mechanism of viperin's broad antiviral activity by demonstrating the protein's ability to synergistically enhance the
José R Peña Cárcamo et al.
Virology, 514, 216-229 (2017-12-05)
Junín arenavirus infections are associated with high levels of interferons in both severe and fatal cases. Upon Junín virus (JUNV) infection a cell signaling cascade initiates, that ultimately attempts to limit viral replication and prevent infection progression through the expression
Ji-Su Jang et al.
Cell death & disease, 9(8), 823-823 (2018-08-03)
Dendritic cells (DCs) are the most potent professional antigen presenting cells and inducers of T cell-mediated immunity. However, few specific markers of mature DCs (mDC) have been reported. A previous microarray analysis revealed expression of mDC-specific genes and identified Rsad2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service