Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU138681

Sigma-Aldrich

MISSION® esiRNA

targeting human MYOG

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTACAGATGCCCACAACCTGCACTCCCTCACCTCCATCGTGGACAGCATCACAGTGGAAGATGTGTCTGTGGCCTTCCCAGATGAAACCATGCCCAACTGAGATTGTCTTCCAAGCCGGGCATCCTTGCGAGCCCCCCAAGCTGGCCACAGATGCCACTACTTCTGTAGCAGGGGCCTCCTAAGCCAGGCTGCCCTGATGCTAGGAAGCCAGCTCTGGGGTGCCATAGGCCAGACTATCCCCTTCCTCATCCATGTAAGGTTAACCCACCCCCCAGCAAGGGACTGGACGCCCTCATTCAGCTGCCTCCTTAGAGGAGAGGGCATCCCCTTTCCAGGGAGGTAAAGCAGGGGACCAGAGCGCCCCCTCGTGTATGCCCCAGCTCAGGGGGCAAACTCAGGAGCTTCCTTTTTATCATAACGCGGCCTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi Yan et al.
International journal of molecular sciences, 16(10), 25014-25030 (2015-10-23)
Fat-induced transcript 1 (FIT1/FITM1) gene is a member of the conserved gene family important for triglyceride-rich lipid droplet accumulation. FIT1 gene displays a similar muscle-specific expression across pigs, mice, and humans. Thus pigs can act as a useful model of
Zhihui Liu et al.
Nature communications, 11(1), 911-911 (2020-02-16)
Embryonal rhabdomyosarcoma (ERMS) is a childhood cancer that expresses myogenic master regulatory factor MYOD but fails to differentiate. Here, we show that the zinc finger transcription factor CASZ1 up-regulates MYOD signature genes and induces skeletal muscle differentiation in normal myoblasts
Adeel Malik et al.
PloS one, 10(7), e0133597-e0133597 (2015-07-23)
Muscle, a multinucleate syncytium formed by the fusion of mononuclear myoblasts, arises from quiescent progenitors (satellite cells) via activation of muscle-specific transcription factors (MyoD, Myf5, myogenin: MYOG, and MRF4). Subsequent to a decline in Pax7, induction in the expression of
Sae-Won Lee et al.
Scientific reports, 5, 16523-16523 (2015-11-14)
Skeletal muscle regeneration occurs continuously to repair muscle damage incurred during normal activity and in chronic disease or injury. Herein, we report that A-kinase anchoring protein 6 (AKAP6) is important for skeletal myoblast differentiation and muscle regeneration. Compared with unstimulated
Majid Rasool Kamli et al.
Biochemical and biophysical research communications, 450(4), 1291-1296 (2014-07-06)
Aldehyde oxidases (AOXs), which catalyze the hydroxylation of heterocycles and oxidation of a wide variety of aldehydic compounds, have been present throughout evolution from bacteria to humans. While humans have only a single functional aldehyde oxidase (AOX1) gene, rodents are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service