Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU136981

Sigma-Aldrich

MISSION® esiRNA

targeting human HDAC4

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGAAGCAGCAGATCCAGAGGCAGATCCTCATCGCTGAGTTCCAGAGGCAGCACGAGCAGCTCTCCCGGCAGCACGAGGCGCAGCTCCACGAGCACATCAAGCAACAACAGGAGATGCTGGCCATGAAGCACCAGCAGGAGCTGCTGGAACACCAGCGGAAGCTGGAGAGGCACCGCCAGGAGCAGGAGCTGGAGAAGCAGCACCGGGAGCAGAAGCTGCAGCAGCTCAAGAACAAGGAGAAGGGCAAAGAGAGTGCCGTGGCCAGCACAGAAGTGAAGATGAAGTTACAAGAATTTGTCCTCAATAAAAAGAAGGCGCTGGCCCACCGGAATCTGAACCACTGCATTTCCAGCGACCCTCGCTACTGGTACGGGAAAACGCAGCACAGTTCCCTTGACCAGAGTTCTCCACCCCAGAGCGGAGTGTCGACCTCCTATAACCACCCGGTCCTGGGAATGTACGACGCCAAAGATGACTTCCCTCTTAGGAAAACAGCTTCTGAACCGAATCTGAAATTACGGTCCAGGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ling X Zhang et al.
American journal of physiology. Cell physiology, 307(4), C358-C372 (2014-06-20)
We have recently shown that in vivo inhibition of histone deacetylase (HDAC) stimulates endogenous myocardial regeneration in infarcted hearts (Zhang L et al. J Pharmacol Exp Ther 341: 285-293, 2012). Furthermore, our observation demonstrates that HDAC inhibition promotes cardiogenesis, which
Todd J Cohen et al.
Molecules and cells, 38(4), 343-348 (2015-03-03)
Fiber type-specific programs controlled by the transcription factor MEF2 dictate muscle functionality. Here, we show that HDAC4, a potent MEF2 inhibitor, is predominantly localized to the nuclei in fast/glycolytic fibers in contrast to the sarcoplasm in slow/oxidative fibers. The cytoplasmic
GuoLiang Zou et al.
Biochimie, 165, 90-99 (2019-05-13)
The cardioprotection of catalpol and its mechanism in diabetic cardiomyopathy (DCM) remains unclear. Here, mouse cardiomyocytes were treated with high glucose (HG) to establish a model of cellular injury induced by HG. In vitro experiments were carried out and confirmed that
Y Yang et al.
Oncogene, 30(19), 2207-2218 (2011-01-19)
The transcriptional activity of the androgen receptor (AR) is regulated by both ligand binding and post-translational modifications, including acetylation and small ubiquitin-like modifier (SUMO)ylation. Histone deacetylases (HDACs) are known to catalyze the removal of acetyl groups from both histones and
Jung-Won Lee et al.
Nature communications, 10(1), 1897-1897 (2019-04-25)
The cellular decision regarding whether to undergo proliferation or death is made at the restriction (R)-point, which is disrupted in nearly all tumors. The identity of the molecular mechanisms that govern the R-point decision is one of the fundamental issues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service