Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU134001

Sigma-Aldrich

MISSION® esiRNA

targeting human TCF7L2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TACCACAGCAAGGTCAACCAGTGTACCCAATCACGACAGGAGGATTCAGACACCCCTACCCCACAGCTCTGACCGTCAATGCTTCCATGTCCAGGTTCCCTCCCCATATGGTCCCACCACATCATACGCTACACACGACGGGCATTCCGCATCCGGCCATAGTCACACCAACAGTCAAACAGGAATCGTCCCAGAGTGATGTCGGCTCACTCCATAGTTCAAAGCATCAGGACTCCAAAAAGGAAGAAGAAAAGAAGAAGCCCCACATAAAGAAACCTCTTAATGCATTCATGTTGTATATGAAGGAAATGAGAGCAAAGGTCGTAGCTGAGTGCACGTTGAAAGAAAGCGCGGCCATCAACCAGATCCTTGGGCGGAGGTGGCATGCACTGTCCAGAGAAGAGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Cui et al.
Aging, 12(2), 1591-1609 (2020-01-24)
Islet β cell mass reduction induced by glucose fluctuation is crucial for the development and progression of T2DM. Chikusetsu saponin IVa (CHS) had protective effects against DM and related injuries. Here we aimed to investigate the role of CHS in
J Wang et al.
British journal of cancer, 111(1), 112-124 (2014-05-31)
Invasion and metastasis remain a critical issue in cervical cancer. However, the underlying mechanism of it in cervical cancer remains unclear. The newly discovered protein, TBLR1, plays a crucial role in regulating various key cellular functions. In this study, western
Chong Chen et al.
Cancer research, 75(8), 1725-1735 (2015-03-07)
Considerable evidence suggests that proinflammatory pathways drive self-renewal of cancer stem-like cells (CSC), but the underlying mechanisms remain mainly undefined. Here we report that the let7 repressor LIN28B and its regulator IKBKB (IKKβ) sustain cancer cell stemness by interacting with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service