Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU132491

Sigma-Aldrich

MISSION® esiRNA

targeting human BTK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGCCATCAAGATGATCAAAGAAGGCTCCATGTCTGAAGATGAATTCATTGAAGAAGCCAAAGTCATGATGAATCTTTCCCATGAGAAGCTGGTGCAGTTGTATGGCGTCTGCACCAAGCAGCGCCCCATCTTCATCATCACTGAGTACATGGCCAATGGCTGCCTCCTGAACTACCTGAGGGAGATGCGCCACCGCTTCCAGACTCAGCAGCTGCTAGAGATGTGCAAGGATGTCTGTGAAGCCATGGAATACCTGGAGTCAAAGCAGTTCCTTCACCGAGACCTGGCAGCTCGAAACTGTTTGGTAAACGATCAAGGAGTTGTTAAAGTATCTGATTTCGGCCTGTCCAGGTATGTCCTGGATGATGAATACACAAGCTCAGTAGGCTCCAAATTTCCAGTCCGGTGGTCCCCACCGGAAGTCCTGATGTATAGCAAGTTCAGCAGCAAATCTGACATTTGGGCTTTTGGGGTTTTGATGTGGGAAATTTACTCCCTGGGGAAGATGCCATATGAGAGATTTACTAACAGTGAGACTGCTGAACACATTGCCCAAGGCCTACGTCTCTACAGGCCTCATCTGGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... BTK(695) , BTK(695)

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chandra Bose Prabaharan et al.
Investigational new drugs, 38(4), 909-921 (2019-08-04)
Treatment response rates to current anticancer therapies for HER2 overexpressing breast cancer are limited and are associated with severe adverse drug reactions. Tyrosine kinases perform crucial roles in cellular processes by mediating cell signalling cascades. Ibrutinib is a recently approved Tyrosine Kinase
E Slinger et al.
Leukemia, 31(12), 2601-2607 (2017-05-04)
The clinical success of B-cell receptor (BCR) signaling pathway inhibitors in chronic lymphocytic leukemia (CLL) is attributed to inhibition of adhesion in and migration towards the lymph node. Proliferation of CLL cells is restricted to this protective niche, but the
Agnieszka Krupa et al.
American journal of physiology. Lung cellular and molecular physiology, 307(6), L435-L448 (2014-08-03)
Previous observations made by our laboratory indicate that Bruton's tyrosine kinase (Btk) may play an important role in the pathophysiology of local inflammation in acute lung injury (ALI)/acute respiratory distress syndrome (ARDS). We have shown that there is cross talk
Genevra Pillinger et al.
Scientific reports, 5, 12949-12949 (2015-08-22)
Approximately 20% of patients with acute myeloid leukaemia (AML) have a mutation in FMS-like-tyrosine-kinase-3 (FLT3). FLT3 is a trans-membrane receptor with a tyrosine kinase domain which, when activated, initiates a cascade of phosphorylated proteins including the SRC family of kinases.
Angela Marina Montalbano et al.
European journal of pharmacology, 736, 35-43 (2014-05-07)
Cigarette smoke extract (CSE) affects the expression of Choline Acetyl-Transferase (ChAT), muscarinic acetylcholine receptors, and mucin production in bronchial epithelial cells. Mucin 5AC (MUC5AC), muscarinic acetylcholine receptor M3, ChAT expression, acetylcholine levels and acetylcholine binding were measured in a human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service