Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU127861

Sigma-Aldrich

MISSION® esiRNA

targeting human EIF2AK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCAGTTTGCCTTCCTGGATTTGTAAATTGTAATGACCTCAAAACTTTAGCAGTTCTTCCATCTGACTCAGGTTTGCTTCTCTGGCGGTCTTCAGAATCAACATCCACACTTCCGTGATTATCTGCGTGCATTTTGGACAAAGCTTCCAACCAGGATACGGGAAGAAGAAATGGCTGGTGATCTTTCAGCAGGTTTCTTCATGGAGGAACTTAATACATACCGTCAGAAGCAGGGAGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tae-Hun Kim et al.
Molecular medicine reports, 16(5), 7585-7590 (2017-09-26)
Kisspeptin is a protein encoded by the KISS1 gene, which has been reported to suppress the metastatic capabilities of various types of cancer cells, through the activation of its G‑protein coupled receptor GPR54. However, the molecular mechanisms underlying the involvement
Jovan Nikolic et al.
PLoS pathogens, 12(10), e1005942-e1005942 (2016-10-18)
Stress granules (SGs) are membrane-less dynamic structures consisting of mRNA and protein aggregates that form rapidly in response to a wide range of environmental cellular stresses and viral infections. They act as storage sites for translationally silenced mRNAs under stress
Stephanie Dabo et al.
Scientific reports, 7(1), 16129-16129 (2017-11-25)
PKR is a cellular kinase involved in the regulation of the integrative stress response (ISR) and pro-inflammatory pathways. Two N-terminal dsRNA Binding Domains (DRBD) are required for activation of PKR, by interaction with either dsRNA or PACT, another cellular DRBD-containing
D C Tanner et al.
Cell death and differentiation, 22(9), 1489-1501 (2015-01-31)
Neuroinflammation associated with degenerative central nervous system disease and injury frequently results in oligodendrocyte death. While promoting oligodendrocyte viability is a major therapeutic goal, little is known about protective signaling strategies. We report that in highly purified rat oligodendrocytes, interferon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service