Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU116291

Sigma-Aldrich

MISSION® esiRNA

targeting human LGALS9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGTGCTCAGAGGTTCCACATCAACCTGTGCTCTGGGAACCACATCGCCTTCCACCTGAACCCCCGTTTTGATGAGAATGCTGTGGTCCGCAACACCCAGATCGACAACTCCTGGGGGTCTGAGGAGCGAAGTCTGCCCCGAAAAATGCCCTTCGTCCGTGGCCAGAGCTTCTCAGTGTGGATCTTGTGTGAAGCTCACTGCCTCAAGGTGGCCGTGGATGGTCAGCACCTGTTTGAATACTACCATCGCCTGAGGAACCTGCCCACCATCAACAGACTGGAAGTGGGGGGCGACATCCAGCTGACCCATGTGCAGACATAGGCGGCTTCCTGGCCCTGGGGCCGGGGGCTGGGGTGTGGGGCAGTCTGGGTCCTCTCATCATCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katrin Schaefer et al.
Glycobiology, 27(9), 878-887 (2017-08-16)
Changes in the T cell surface redox environment regulate critical cell functions, such as cell migration, viral entry and cytokine production. Cell surface protein disulfide isomerase (PDI) contributes to the regulation of T cell surface redox status. Cell surface PDI
Tomokazu Nunoue et al.
Scientific reports, 11(1), 5991-5991 (2021-03-18)
The adipose tissue is regarded as an endocrine organ and secretes bioactive adipokines modulating chronic inflammation and oxidative stress in obesity. Gal-9 is secreted out upon cell injuries, interacts with T-cell immunoglobulin-3 (Tim-3) and induces apoptosis in activated Th1 cells.
Akira Nishio et al.
Hepatology (Baltimore, Md.), 65(1), 18-31 (2016-09-20)
Natural killer (NK) cell activation is associated with both liver injury and persistent infection in chronic hepatitis C (CHC); however, the detailed mechanism of this activation has not yet been fully elucidated. Because galectin-9 (Gal-9) has been reported to be
Wenxi Zhou et al.
Biomaterials, 268, 120546-120546 (2020-12-01)
Immunotherapy has gained increasing focus in treating pancreatic ductal adenocarcinoma (PDAC), since conventional therapies like chemotherapy could not provide satisfactory improvement in overall survival outcome of PDAC patients. However, it is still not the game changing solution due to the
Ran Lv et al.
Molecular medicine reports, 16(6), 9111-9119 (2017-10-11)
Generally considered as a potent pro‑inflammatory signal, β‑galactosidelectin suppresses T cell receptor activation, can both promote and inhibit integrin‑mediated adhesion and is required in nuclear pre‑mRNA splicing. Galectin‑9 (Gal‑9), a member of β‑galactoside lectin, is involved many processes of T cell‑mediated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service