Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU111891

Sigma-Aldrich

MISSION® esiRNA

targeting human NANOG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTCCTTCCTCCATGGATCTGCTTATTCAGGACAGCCCTGATTCTTCCACCAGTCCCAAAGGCAAACAACCCACTTCTGCAGAGAAGAGTGTCGCAAAAAAGGAAGACAAGGTCCCGGTCAAGAAACAGAAGACCAGAACTGTGTTCTCTTCCACCCAGCTGTGTGTACTCAATGATAGATTTCAGAGACAGAAATACCTCAGCCTCCAGCAGATGCAAGAACTCTCCAACATCCTGAACCTCAGCTACAAACAGGTGAAGACCTGGTTCCAGAACCAGAGAATGAAATCTAAGAGGTGGCAGAAAAACAACTGGCCGAAGAATAGCAATGGTGTGACGCAGAAGGCCTCAGCACCTACCTACCCCAGCCTTTACTCTTCCTACCACCAGGGATGCCTGGTGAACCCGACTGGGAACCTTCCAATGTGGAGCAACCAGACCTGGAACAAT

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Li et al.
Oncology letters, 17(1), 555-563 (2019-01-19)
NANOGP8 is one of the NANOG pseudogenes and is expressed together with NANOG in multiple tumor tissues and cell lines. The biological functions of NANOGP8 in progression of gastric cancer are unclear. In the present study, the role of NANOGP8
Yuki Katsura et al.
Cancers, 11(2) (2019-02-06)
Excess iron causes cancer and is thought to be related to carcinogenesis and cancer progression including stemness, but the details remain unclear. Here, we hypothesized that stemness in cancer is related to iron metabolism and that regulating iron metabolism in
Khalid Arif et al.
OncoTargets and therapy, 8, 1327-1334 (2015-06-18)
There is an accumulation of evidence that shows a significant role of cancer stem cells in tumor initiation, proliferation, relapse, and metastasis. Nanog is the most important core transcription marker of stem cells, known by its role in maintaining pluripotency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service