Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU108371

Sigma-Aldrich

MISSION® esiRNA

targeting human ACTR3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTCAAAGGAGGGTGATGAAAGGTGTTGATGACCTAGACTTCTTCATTGGTGATGAAGCAATAGAAAAACCTACATATGCAACAAAGTGGCCAATCCGCCATGGTATAGTTGAAGATTGGGACTTAATGGAAAGGTTTATGGAGCAAGTGATCTTTAAATATTTAAGGGCAGAACCTGAAGACCATTATTTTCTTTTGACTGAACCTCCATTGAATACTCCAGAAAACAGGGAATATACTGCTGAAATAATGTTTGAGTCCTTCAATGTTCCAGGCTTGTACATTGCTGTGCAGGCTGTTCTTGCCTTAGCTGCATCTTGGACCTCAAGACAAGTAGGAGAACGGACGTTGACCGGTACGGTAATAGACAGTGGAGATGGTGTCACTCATGTCATTCCTGTGGCTGAAGGGTATGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandra S Chikina et al.
Biology of the cell, 111(10), 245-261 (2019-08-14)
Metastatic disease is caused by the ability of cancer cells to reach distant organs and form secondary lesions at new locations. Dissemination of cancer cells depends on their migration plasticity - an ability to switch between motility modes driven by
Meraj H Khan et al.
Journal of cell science, 130(18), 3094-3107 (2017-08-05)
Sharpin, a multifunctional adaptor protein, regulates several signalling pathways. For example, Sharpin enhances signal-induced NF-κB signalling as part of the linear ubiquitin assembly complex (LUBAC) and inhibits integrins, the T cell receptor, caspase 1 and PTEN. However, despite recent insights
Daniel L Galvan et al.
The Journal of clinical investigation, 129(7), 2807-2823 (2019-05-08)
Phosphorylation of Dynamin-related protein1 (Drp1) represents an important regulatory mechanism for mitochondrial fission. Here we established the role of Drp1 Serine 600 (S600) phosphorylation on mitochondrial fission in vivo, and assessed the functional consequences of targeted elimination of the Drp1S600

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service