Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU106991

Sigma-Aldrich

MISSION® esiRNA

targeting human UQCRFS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAAATTGAGCAGGAAGCTGCAGTTGAATTATCACAGTTGAGGGACCCACAGCATGATCTAGATCGAGTAAAGAAACCTGAATGGGTTATCCTGATAGGTGTTTGCACTCATCTTGGCTGTGTACCCATTGCAAATGCAGGAGATTTTGGTGGTTATTACTGCCCTTGCCATGGGTCACACTATGATGCATCTGGCAGGATCAGATTGGGTCCTGCTCCTCTCAACCTTGAAGTCCCCACGTATGAGTTCACCAGTGACGATATGGTGATTGTTGGTTAAGAGACTTGGACTCAAGTCATAGGCTTCTTTCAGTCTTTATGTCACCTCAGGAGACTTATTTGAGAGGAAGCCTTCTGTACTTGAAGTTGATTTGAAATATGTAAGAATTGATGATGTATTTGCAAACATTAATGTGAAATAAATTGAATTTAATGTTGAATACTTTCAGGCATTCACTTAATAAAGACACTGTTAAGCACTGTTATGCTCAGTCATACACGCGAAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao Yang et al.
Antioxidants & redox signaling, 32(7), 447-462 (2019-08-29)
Aims: It is known that mitochondrial reactive oxygen species generation ([ROS]m) causes the release of Ca2+via ryanodine receptor-2 (RyR2)
Lin Mei et al.
Nature communications, 11(1), 3527-3527 (2020-07-17)
Ca2+ signaling in pulmonary arterial smooth muscle cells (PASMCs) plays an important role in pulmonary hypertension (PH). However, the underlying specific ion channel mechanisms remain largely unknown. Here, we report ryanodine receptor (RyR) channel activity and Ca2+ release both are
Yoshiko Kaku et al.
Cellular signalling, 27(9), 1713-1719 (2015-05-26)
The present study investigated 1,2-diarachidonoyl-sn-glycero-3-phosphoethanolamine (DAPE)-induced cell death in malignant pleural mesothelioma (MPM) cells. DAPE reduced cell viability in NCI-H28, NCI-H2052, NCI-H2452, and MSTO-211H MPM cell lines in a concentration (1-100μM)-dependent manner. In the flow cytometry using propidium iodide (PI)
W He et al.
Oncogene, 33(23), 3004-3013 (2013-07-09)
Killing cancer cells through the induction of apoptosis is one of the main mechanisms of chemotherapy. However, numerous cancer cells have primary or acquired apoptosis resistance, resulting in chemoresistance. In this study, using a novel chalcone derivative chalcone-24 (Chal-24), we
H Schoeneberger et al.
Oncogene, 34(31), 4032-4043 (2014-11-11)
Evasion of apoptosis in pediatric acute lymphoblastic leukemia (ALL) is linked to aberrant expression of inhibitor of apoptosis (IAP) proteins and dysregulated redox homeostasis, rendering leukemic cells vulnerable to redox-targeting therapies. Here we discover that inhibition of antioxidant defenses via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service