Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU106621

Sigma-Aldrich

MISSION® esiRNA

targeting human SUMO1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCAAAGACAGGGTGTTCCAATGAATTCACTCAGGTTTCTCTTTGAGGGTCAGAGAATTGCTGATAATCATACTCCAAAAGAACTGGGAATGGAGGAAGAAGATGTGATTGAAGTTTATCAGGAACAAACGGGGGGTCATTCAACAGTTTAGATATTCTTTTTATTTTTTTTCTTTTCCCTCAATCCTTTTTTATTTTTAAAAATAGTTCTTTTGTAATGTGGTGTTCAAAACGGAATTGAAAACTGGCACCCCATCTCTTTGAAACATCTGGTAATTTGAATTCTAGTGCTCATTATTCATTATTGTTTGTTTTCATTGTGCTGATTTTTGGTGATCAAGCCTCAGTCCCCTTCATATTACCCTCTCCTTTTTAAAAATTACGTGTGCACAGAGAGGTCACCTTTTTCAGGACATTGCATTTTCAGGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yufeng Yao et al.
Pulmonary pharmacology & therapeutics, 55, 38-49 (2019-02-01)
Pulmonary arterial hypertension (PAH) is a life-threatening disease without effective therapies. PAH is associated with a progressive increase in pulmonary vascular resistance and irreversible pulmonary vascular remodeling. SUMO1 (small ubiquitin-related modifier 1) can bind to target proteins and lead to
Sabine Hergovits et al.
Journal of cellular and molecular medicine, 21(11), 3087-3099 (2017-06-01)
Interleukin (IL)-6-type cytokines have no direct antiviral activity; nevertheless, they display immune-modulatory functions. Oncostatin M (OSM), a member of the IL-6 family, has recently been shown to induce a distinct number of classical interferon stimulated genes (ISG). Most of them
Lin-Nan Zhu et al.
Frontiers in cellular neuroscience, 12, 262-262 (2018-09-11)
Methamphetamine (METH) is an illegal and widely abused psychoactive stimulant. METH abusers are at high risk of neurodegenerative disorders, including Parkinson's disease (PD). Previous studies have demonstrated that METH causes alpha-synuclein (α-syn) aggregation in the both laboratory animal and human.
Dao-Ying Yuan et al.
Oncology reports, 38(4), 2360-2368 (2017-08-10)
Radiotherapy is one of the most effective non-surgical treatments for oral squamous cell carcinoma. However, radioresistance remains a major impediment to radiotherapy. Although BetA (Betulinic acid) can induce radiosensitization, the underlying mechanism and whether it could induce radiosensitization in oral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service