Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU104101

Sigma-Aldrich

MISSION® esiRNA

targeting human ATXN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAGGAACTTGAGGCTTTGGAAAATGACGTATCTAATGGATGGGATCCCAATGATATGTTTCGATATAATGAAGAAAATTATGGTGTAGTGTCTACGTATGATAGCAGTTTATCTTCGTATACAGTGCCCTTAGAAAGAGATAACTCAGAAGAATTTTTAAAACGGGAAGCAAGGGCAAACCAGTTAGCAGAAGAAATTGAGTCAAGTGCCCAGTACAAAGCTCGAGTGGCCCTGGAAAATGATGATAGGAGTGAGGAAGAAAAATACACAGCAGTTCAGAGAAATTCCAGTGAACGTGAGGGGCACAGCATAAACACTAGGGAAAATAAATATATTCCTCCTGGACAAAGAAATAGAGAAGTCATATCCTGGGGAAGTGGGAGACAGAATTCACCGCGTATGGGCCAGCCTGGATCGGGCTCCATGCCATCAAGATCCACTTCTCACACTTCAGATTTCAACCCGAATTCTGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jongbo Lee et al.
PLoS biology, 18(12), e3001002-e3001002 (2020-12-29)
Nucleocytoplasmic transport (NCT) defects have been implicated in neurodegenerative diseases such as C9ORF72-associated amyotrophic lateral sclerosis and frontotemporal dementia (C9-ALS/FTD). Here, we identify a neuroprotective pathway of like-Sm protein 12 (LSM12) and exchange protein directly activated by cyclic AMP 1
Nesli Ece Sen et al.
Neurobiology of disease, 96, 115-126 (2016-10-19)
Ataxin-2 (ATXN2) polyglutamine domain expansions of large size result in an autosomal dominantly inherited multi-system-atrophy of the nervous system named spinocerebellar ataxia type 2 (SCA2), while expansions of intermediate size act as polygenic risk factors for motor neuron disease (ALS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service