Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU099501

Sigma-Aldrich

MISSION® esiRNA

targeting human F11R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCTTTCCTACCACTGCTGAGTGGCCTGGAACTTGTTTAAAGTGTTTATTCCCCATTTCTTTGAGGGATCAGGAAGGAATCCTGGGTATGCCATTGACTTCCCTTCTAAGTAGACAGCAAAAATGGCGGGGGTCGCAGGAATCTGCACTCAACTGCCCACCTGGCTGGCAGGGATCTTTGAATAGGTATCTTGAGCTTGGTTCTGGGCTCTTTCCTTGTGTACTGACGACCAGGGCCAGCTGTTCTAGAGCGGGAATTAGAGGCTAGAGCGGCTGAAATGGTTGTTTGGTGATGACACTGGGGTCCTTCCATCTCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonas Franz et al.
PloS one, 11(1), e0146598-e0146598 (2016-01-06)
Endothelial barriers have a central role in inflammation as they allow or deny the passage of leukocytes from the vasculature into the tissue. To bind leukocytes, endothelial cells form adhesive clusters containing tetraspanins and ICAM-1, so-called endothelial adhesive platforms (EAPs).
Min Xie et al.
Cancer letters, 443, 67-79 (2018-12-07)
Multiple studies have revealed that long non-coding RNAs (lncRNAs) extensively participate in human cancer malignant progression. The long intergenic non-protein coding RNA 707 (LINC00707), 3087 bp in length, was recently reported to be an essential oncogene in promoting lung adenocarcinoma
Rodrigo G B Cruz et al.
Cancers, 13(4) (2021-03-07)
The success of breast cancer therapies targeting the human epidermal growth factor receptor-2 (HER2) is limited by the development of drug resistance by mechanisms including upregulation of HER3. Having reported that HER2 expression and resistance to HER2-targeted therapies can be
Nikola Sladojevic et al.
Neurobiology of disease, 67, 57-70 (2014-03-25)
Proinflammatory mediators trigger intensive postischemic inflammatory remodeling of the blood-brain barrier (BBB) including extensive brain endothelial cell surface and junctional complex changes. Junctional adhesion molecule-A (JAM-A) is a component of the brain endothelial junctional complex with dual roles: paracellular route
Hüseyin Tuncay et al.
Nature communications, 6, 8128-8128 (2015-08-27)
Planar spindle orientation in polarized epithelial cells depends on the precise localization of the dynein-dynactin motor protein complex at the lateral cortex. The contribution of cell adhesion molecules to the cortical localization of the dynein-dynactin complex is poorly understood. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service