Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU092681

Sigma-Aldrich

MISSION® esiRNA

targeting human ADK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGGTACAAGGGTATGGTAATGCTTGTAGAATCTTTATTATCTCAACAATCTAAAAAATGATGTTTATTTCCATAGTTTGATAGTGCCACTTAAATGCCAATTAAACAAGAATATAACATTTCAATAGAAATTTTTATTTCATTTTCAATTACTTTGTAAATTCGTGTGTATTTAGTACACTGATTTGTTTTTTTACATTTCTGCTTTGAATGCAGATGCAATTTAATATAATAGATTTTTTAATGAATTAATCTTAACATAGTAATCTTTAGCTTTTTATACAAATATATTTAATTTAGGAGTATATGTGTGTCTATACACACACATACATAAATATACCACATATACACCTGATAGTCAAATAAGGTACAGAAATTTTATCTTGTCAATTATGCCAAATAATCTCTTTAATGTGCACTCAAACA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... ADK(132) , ADK(132)

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Jin et al.
Journal of molecular histology, 47(3), 259-271 (2016-03-18)
Adenosine kinase (ADK) plays a pivotal role in regulating brain function by regulating adenosine level, and ADK inhibition protects against neuronal damage in cerebral ischemia and epilepsy; however, the effects of ADK in traumatic brain injury (TBI) have not been
Iwona Pelikant-Malecka et al.
The international journal of biochemistry & cell biology, 88, 31-43 (2017-03-23)
4-pirydone-3-carboxamide-1β-d-ribonucleoside (4PYR) is an endogenous nucleoside that could be converted to triphosphates, diphosphates, monophosphates and an analogue of NAD - 4PYRAD. Elevated level of these compounds have been reported in chronic renal failure, cancer and active HIV infection. However, little
Alexander J Valvezan et al.
JCI insight, 5(7) (2020-04-10)
Recent studies in distinct preclinical tumor models have established the nucleotide synthesis enzyme inosine-5'-monophosphate dehydrogenase (IMPDH) as a viable target for antitumor therapy. IMPDH inhibitors have been used clinically for decades as safe and effective immunosuppressants. However, the potential to
Wei Cao et al.
Life sciences, 256, 117972-117972 (2020-06-17)
Acute kidney injury (AKI) has a high morbidity and mortality, and there is no targeted treatment yet. One of the main causes of AKI is ischemia-reperfusion (IR). Increased release of adenosine under stress and hypoxia exerts anti-inflammatory and antioxidant effects.
Tal Israeli et al.
Journal of cell science, 131(15) (2018-07-14)
AMPK-mTORC1 signaling senses nutrient availability, thereby regulating autophagy. Surprisingly, we found that, in β-cells, the AMPK activator 5-amino-4-imidazolecarboxamide ribofuranoside (AICAR) inhibited, rather than stimulated, autophagy. AICAR is an intermediate in the generation of inosine monophosphate, with subsequent conversion to other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service