Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU091361

Sigma-Aldrich

MISSION® esiRNA

targeting human MRE11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCTCTGTAAAAAGATCCCTGAGATTATTCCTTCTTCTAGTTTTATGCGACAGCTTTACTTTAAAATTCAAGTTATACATCTTGGGAGTACAATGGCCCGACATTTCTTCATAGGTAGAAACAAATACTTGACTCAGTGATACTCATGACCATTAGAATAGTCATACCTGGAATGTGTCAAATTATAAGAGACAGACACTTGGTTAGTGGCTGCCTCATATAGCACTTTTGAAGAGGCCTAAGTCAAAACTTGCAATATAACATTCTATTGACTTTCTTAAAAATATTTTTTCTGTACCTAACTTGAGCATAAGGGTTATTTGAGCAAGTAACATTAACTCAGTGGAAGGCATTGTCCTGTGAAATATTCTTAGGCAGATCTGCCCACATCTTTATTGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Biochem/physiol Actions

MRE11A (meiotic recombination 11 homolog A) is a nuclease which forms complex with Rad50 (DNA repair protein) and Nbs1 (nijmegen breakage syndrome protein 1). This complex works as a sensor of DNA double strand breaks, and is involved in DNA repair processes and DNA damage response. Absence of the complex activity causes developmental and/or degenerative neuronal disorders. In homologous recombination repair, MRE11A is responsible for the 3′-to-5′ exonuclease activity, leading to the generation of protruding 3′ ssDNA at double strand breaks. MRE11A is also an oncoprotein which is associated with colorectal cancer and malignant breast cancer. Hypomorphic mutations in this gene leads to Ataxia-Telangiectasia like disorder.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of MRE11 and Clinicopathologic Characteristics in Recurrence of Breast Cancer: Individual and Cumulated Receiver Operating Characteristic Analyses.
Yang CH
BioMed Research International, 2017, 2563910-2563910 (2017)
Rad51 recombinase prevents Mre11 nuclease-dependent degradation and excessive PrimPol-mediated elongation of nascent DNA after UV irradiation.
Vallerga MB
Proceedings of the National Academy of Sciences of the USA, 112, E6624-E6624 (2015)
M Petroni et al.
Cell death and differentiation, 23(2), 197-206 (2015-06-13)
The MRE11/RAD50/NBS1 (MRN) complex is a major sensor of DNA double strand breaks, whose role in controlling faithful DNA replication and preventing replication stress is also emerging. Inactivation of the MRN complex invariably leads to developmental and/or degenerative neuronal defects
A Bakr et al.
Nucleic acids research, 43(6), 3154-3166 (2015-03-11)
Ataxia-telangiectasia mutated (ATM) is needed for the initiation of the double-strand break (DSB) repair by homologous recombination (HR). ATM triggers DSB end resection by stimulating the nucleolytic activity of CtIP and MRE11 to generate 3'-ssDNA overhangs, followed by RPA loading
Ying Wai Chan et al.
Nature cell biology, 20(1), 92-103 (2017-12-20)
The resolution of joint molecules that link recombining sister chromatids is essential for chromosome segregation. Here, we determine the fate of unresolved recombination intermediates arising in cells lacking two nucleases required for resolution (GEN1 -/- knockout cells depleted of MUS81).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service