Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU088291

Sigma-Aldrich

MISSION® esiRNA

targeting human RASSF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGCAAACAGAACAGGGTCATGTTTGCAGGGGTGACGGCCCTCATCTATGAGGAAAGGTTTTGGATCTTGAATGTGGTCTCAGGATATCCTTATCAGAGCTAAGGGTGGGTGCTCAGAATAAGGCAGGCATTGAGGAAGAGTCTTGGTTTCTCTCTACAGTGCCAACTCCTCACACACCCTGAGGTCAGGGAGTGCTGGCTCACAGTACAGCATGTGCCTTAATGCTTCATATGAGGAGGATGTCCCTGGGCCAGGGTCTGTGTGAATGTGGGCACTGGCCCAGGTTCATACCTTATTTGCTAATCAAAGCCAGGGTCTCTCCCTCAGGTGTTTTTTATGAAGTGCGTGAATGTATGTAATGTGTGGTGGCCTCAGCTGAATGCCTCCTGTGGGGAAAGGGGTTGGGGTGACAGTCATCATCAGGGCCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junhui Yu et al.
Oncology reports, 40(4), 1959-1970 (2018-08-15)
MicroRNA (miR)‑181a is a member of the miR‑181 family that serves a key role in the pathogenesis of various cancer types. The present study aimed to investigate the interaction between miR‑181a and Ras association domain family protein1 isoform A (RASSF1A)
Swati Dabral et al.
Nature communications, 10(1), 2130-2130 (2019-05-16)
Hypoxia signaling plays a major role in non-malignant and malignant hyperproliferative diseases. Pulmonary hypertension (PH), a hypoxia-driven vascular disease, is characterized by a glycolytic switch similar to the Warburg effect in cancer. Ras association domain family 1A (RASSF1A) is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service