Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU085901

Sigma-Aldrich

MISSION® esiRNA

targeting human GLS (2)

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAAAAGAGCCGAGTGGACTAAGATTCAACAAACTATTTTTGAATGAAGATGATAAACCACATAATCCTATGGTAAATGCTGGAGCAATTGTTGTGACTTCACTAATAAAGCAAGGAGTAAATAATGCTGAAAAATTTGACTATGTCATGCAGTTTTTGAATAAGATGGCTGGTAATGAATATGTTGGATTCAGTAATGCAACGTTTCAGTCTGAAAGAGAAAGTGGAGATCGAAATTTTGCAATAGGATATTACTTAAAAGAAAAGAAGTGTTTTCCAGAAGGCACAGACATGGTTGGTATATTAGACTTCTACTTCCAGCTGTGCTCCATTGAAGTGACTTGTGAATCAGCCAGTGTGATGGCTGCGACACTGGCTAATGGTGGTTTCTGCCCAATTACTGGTGAAAGAGTACTGAGCCCTGAAGCAGTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denis A Mogilenko et al.
Cell, 177(5), 1201-1216 (2019-04-30)
Innate immune responses are intricately linked with intracellular metabolism of myeloid cells. Toll-like receptor (TLR) stimulation shifts intracellular metabolism toward glycolysis, while anti-inflammatory signals depend on enhanced mitochondrial respiration. How exogenous metabolic signals affect the immune response is unknown. We
Jae-Seon Lee et al.
Biochemical and biophysical research communications, 477(3), 374-382 (2016-06-25)
We found that non-small cell lung cancer (NSCLC) is remarkably sensitive to the regulation of glutamine supply by testing the metabolic dependency of 11 cancer cell lines against regulation of glycolysis, autophagy, fatty acid synthesis, and glutamine supply. Glutamine is
Lisha Xiang et al.
Cell death & disease, 10(2), 40-40 (2019-01-25)
Cancer cells re-program their metabolic machinery to meet the requirements of malignant transformation and progression. Glutaminase 1 (GLS1) was traditionally known as a mitochondrial enzyme that hydrolyzes glutamine into glutamate and fuels rapid proliferation of cancer cells. However, emerging evidence
Xuan Qu et al.
Biochemical and biophysical research communications, 504(2), 415-421 (2018-08-15)
Oncogenic c-Myc-induced metabolic reprogramming triggers cellular dependency on exogenous glucose and glutamine. Understanding how nutrients are used may provide new target for therapeutic intervention. We previously provided an alternate route to c-Myc-driven glucose metabolism via the repression of thioredoxin-interacting protein
Yuta Mizuno et al.
Oncology letters, 19(4), 2934-2942 (2020-03-29)
The high expression of metabolic enzymes, including glutaminase (GA) and lactate dehydrogenase A (LDHA), which contribute to bioenergetics and biosynthesis of mammalian cells, has been identified in a variety of cancer types. The current study indicated intratumoral heterogeneity with respect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service