Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU085201

Sigma-Aldrich

MISSION® esiRNA

targeting human NDRG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTCGAGGCTAGAGGCATTTGGAACAACAAATCTACGTAGTTAACTTGAAGAAACCGATTTTTAAAGTTGGTGCATCTAGAAAGCTTTGAATGCAGAAGCAAACAAGCTTGATTTTTCTAGCATCCTCTTAATGTGCAGCAAAAGCAGGCGACAAAATCTCCTGGCTTTACAGACAAAAATATTTCAGCAAACGTTGGGCATCATGGTTTTTGAAGGCTTTAGTTCTGCTTTCTGCCTCTCCTCCACAGCCCCAACCTCCCACCCCTGATACATGAGCCAGTGATTATTCTTGTTCAGGGAGAAGATCATTTAGATTTGTTTTGCATTCCTTAGAATGGAGGGCAACATTCCACAGCTGCCCTGGCTGTGATGAGTGTCCTTGCAGGGGCCGGAGTAGGAGCACTGGGGTGGGGGTGGAATTGGGGTTACTCGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Cen et al.
Oncology reports, 37(2), 1189-1195 (2017-01-12)
The N-myc downstream regulated gene 1 (NDRG1) is differently expressed in human malignancies according to the tumor type. We investigated the expression of NDRG1 in pancreatic cancer tissues and cell lines as well as how it affects tumor growth, invasion and
Jinju Liu et al.
Human cell, 33(4), 1176-1185 (2020-08-07)
Numerous studies demonstrated that microRNAs (miRNAs) were highly involved in pancreatic cancer development. However, the functional roles of many miRNAs remain elusive in pancreatic cancer. In the present study, we analyzed previous published microarray data and found that miR-1469-5p was
Nan Meng et al.
Molecular reproduction and development, 86(9), 1210-1223 (2019-07-25)
Embryo implantation is an essential step for a successful pregnancy, and any defect in this process can lead to a range of pregnancy pathologies. The objective of this study was to explore the role of N-myc downregulated gene 1 (NDRG1)
Aiwei Li et al.
Scientific reports, 9(1), 5166-5166 (2019-03-28)
N-myc downstream regulated gene 1 (NDRG1) is an intracellular protein involved in cell differentiation and was recently reported to exert various effects in several cancers. However, its expression and role in bladder cancer remain unclear. Our study enrolled 100 bladder
Zhi-Yan Hu et al.
Biochimica et biophysica acta, 1852(9), 1876-1886 (2015-06-15)
N-myc downstream-regulated gene 1 (NDRG1) has been implicated in tumorigenesis and metastasis in different cancers. However, its role in nasopharyngeal carcinoma remains unknown. We found that NDRG1 expression level was high in nasopharyngeal cancer 5-8F cells but low in 5-8F-LN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service