Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU075501

Sigma-Aldrich

MISSION® esiRNA

targeting human CUX1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGCAAGGAAGCTGAAGCAGCTTTCTTGAATGTCTACAAAAGATTGATTGACGTCCCAGATCCCGTACCAGCTTTGGATCTCGGACAGCAACTCCAGCTCAAAGTGCAGCGCCTGCACGATATTGAAACAGAGAACCAGAAACTTAGGGAAACTCTGGAAGAATACAACAAGGAATTTGCTGAAGTGAAAAATCAAGAGGTTACGATAAAAGCACTTAAAGAGAAAATCCGAGAATATGAACAGACACTGAAGAACCAAGCCGAAACCATAGCTCTTGAGAAGGAACAGAAGTTACAGAATGACTTTGCAGAAAAGGAGAGAAAGCTGCAGGAGACACAGATGTCCACCACCTCAAAGCTGGAGGAAGCTGAGCATAAGGTTCAGAGCCTACAAACAGCCCTGGAAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liza J Burton et al.
PloS one, 14(4), e0214844-e0214844 (2019-04-10)
Triple-Negative Breast Cancers (TNBCs) are the most difficult to treat subtype of breast cancer and are often associated with high nuclear expression of Snail and Cathepsin L (Cat L) protease. We have previously shown that Snail can increase Cat L
Xiao Zhang et al.
Neurochemical research, 45(12), 2840-2855 (2020-10-02)
Circular RNAs (circRNAs) played pivotal roles in the initiation and progression of cancers. CircRNA cut like homeobox 1 (circ-CUX1; hsa_circ_0132813) has been reported to contribute to neuroblastoma (NB) development by previous study. Furthermore, previous works reported that microRNA-16-5p (miR-16-5p) was
Junfeng Wang et al.
Oncology reports, 37(5), 3068-3074 (2017-04-14)
The homeobox transcription factor CUTL1 has been associated with cellular proliferation and cell cycle progression, and CUTL1 functions as an oncogene. The aim of the present study was to investigate whether CUTL1 participates in epithelial-mesenchymal transition (EMT). The expression levels
Tian Gao et al.
Cancer biology & medicine, 17(2), 371-386 (2020-06-27)
Objective: Osteosarcoma is a common primary highly malignant bone tumor. Kinesin family member 18B (KIF18B) has been identified as a potential oncogene involved in the development and metastasis of several cancer types. While KIF18B overexpression in osteosarcoma tissue is clearly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service