Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU073721

Sigma-Aldrich

MISSION® esiRNA

targeting human TRPV1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAGGACAAGTGGGACAGATTCGTCAAGCGCATCTTCTACTTCAACTTCCTGGTCTACTGCCTGTACATGATCATCTTCACCATGGCTGCCTACTACAGGCCCGTGGATGGCTTGCCTCCCTTTAAGATGGAAAAAACTGGAGACTATTTCCGAGTTACTGGAGAGATCCTGTCTGTGTTAGGAGGAGTCTACTTCTTTTTCCGAGGGATTCAGTATTTCCTGCAGAGGCGGCCGTCGATGAAGACCCTGTTTGTGGACAGCTACAGTGAGATGCTTTTCTTTCTGCAGTCACTGTTCATGCTGGCCACCGTGGTGCTGTACTTCAGCCACCTCAAGGAGTATGTGGCTTCCATGGTATTCTCCCTGGCCTTGGGCTGGACCAACATGCTCTACTACACCCGCGGTTTC

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gwan Hee Han et al.
Cancer genomics & proteomics, 17(3), 309-319 (2020-04-30)
Transient receptor potential vanilloid type 1 (TRPV1) has been studied in human malignancies, but has not been studied in epithelial ovarian cancer (EOC). We, therefore, investigated the significance of TRPV1 and correlation with phosphatase and tension homolog (PTEN) in EOC.
Ayumi Maeda et al.
Molecular nutrition & food research, 62(11), e1800086-e1800086 (2018-04-24)
The prevalence of type 2 diabetes mellitus (T2DM) is increasing yearly worldwide. Glycemic control is the basis for the treatment of T2DM, as it can prevent the progress of associated complications. Spices possess various health beneficial effects on humans. The
Mingming Ren et al.
Cardiovascular therapeutics, 34(6), 482-488 (2016-09-24)
Accumulating evidence showed that transient receptor potential channels play an important role in the regulation of cardiomyocyte differentiation. The vanilloid receptor 1 (VR1) is a member of the transient receptor channel super family and is expressed in cardiomyocytes. However, its
Masayoshi Kawase et al.
Scientific reports, 10(1), 9687-9687 (2020-06-18)
Despite successful clinical application of non-equilibrium atmospheric pressure plasma (APP), the details of the molecular mechanisms underlying APP-inducible biological responses remain ill-defined. We previously reported that exposure of 3T3L1 cells to APP-irradiated buffer raised the cytoplasmic free Ca2+ ([Ca2+]i) concentration
Shuo Wang et al.
Channels (Austin, Tex.), 14(1), 59-68 (2020-02-23)
The natural outcome of abdominal aortic aneurysm (AAA) is that of slow progression and ultimate rupture, then a life-threatening hemorrhage consequently. Ruptured AAA is a dramatic catastrophe and constitutes one of the leading causes of acute death in elderly men.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service