Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU072991

Sigma-Aldrich

MISSION® esiRNA

targeting human HMGB1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACACCCTGCATATCATAATGGGGGTAAAGTTAAGTTGAGATAGTTTTCATCCATAACTGAACATCCAAAATCTTGATCAGTTAAGAAATTTCACATAGCCCACTTACATTTACAAACTGAAGAGTAATCAATCTACTCAAAGCATGGGATTATTAGAATCAAACATTTTGAAAGTCTGTCCTTGAAGGACTAATAGAAAAGTATGTTCTAACCTTTACATGAGGACTCTATTCTTTAACTCCCATTACCATGTAATGGCAGTTATATTTTGCAGTTCCCACATTAAAGAAGACCTGAGAATGTATCCCCAAAAGCGTGAGCTTAAAATACAAGACTGCCATATTAAATTTTTTGTTGACATTAGTCTCAGTGAAGACTATGAAAATGCTGGCTATAGATGTCTTTTCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Zhang et al.
Journal of neuroinflammation, 12, 156-156 (2015-09-05)
Mounting evidence has indicated that high-mobility group box 1 (HMGB1) is involved in cell activation and migration. Our previous study demonstrated that methamphetamine mediates activation of astrocytes via sigma-1 receptor (σ-1R). However, the elements downstream of σ-1R in this process
Yan Chen et al.
International journal of clinical and experimental pathology, 8(6), 6683-6691 (2015-08-12)
Diabetic nephropathy (DN) is one of the most devastating complications of diabetes, leading the cause of end-stage renal disease (ESRD). And investigations into mechanisms underlying renal inflammation may provide new insight into novel therapeutic targets for patients with DN. However
Minghua Yang et al.
Autophagy, 11(2), 214-224 (2015-01-22)
Both apoptosis ("self-killing") and autophagy ("self-eating") are evolutionarily conserved processes, and their crosstalk influences anticancer drug sensitivity and cell death. However, the underlying mechanism remains unclear. Here, we demonstrated that HMGB1 (high mobility group box 1), normally a nuclear protein
Zhe Liu et al.
Chinese journal of cancer research = Chung-kuo yen cheng yen chiu, 27(3), 267-278 (2015-07-15)
The purpose of this study was to examine the effect of gemcitabine (GEM) on microRNA-218 (miR-218) expression in human pancreatic cancer cells. Quantitative reverse transcription polymerase chain reaction (qRT-PCR) was performed to examine the differences in miR-218 expression between the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service