Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU071471

Sigma-Aldrich

MISSION® esiRNA

targeting human AL513122.2, GSN, AL513122.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCACCAACCTTTATGGAGACTTCTTCACGGGCGACGCCTACGTCATCCTGAAGACAGTGCAGCTGAGGAACGGAAATCTGCAGTATGACCTCCACTACTGGCTGGGCAATGAGTGCAGCCAGGATGAGAGCGGGGCGGCCGCCATCTTTACCGTGCAGCTGGATGACTACCTGAACGGCCGGGCCGTGCAGCACCGTGAGGTCCAGGGCTTCGAGTCGGCCACCTTCCTAGGCTACTTCAAGTCTGGCCTGAAGTACAAGAAAGGAGGTGTGGCATCAGGATTCAAGCACGTGGTACCCAACGAGGTGGTGGTGCAGAGACTCTTCCAGGTCAAAGGGCGGCGTGTGGTCCGTGCCACCGAGGTACCTGTGTCCTGGGAGAGCTTCAACAATGGCGACTGCTTCATCCTGGACCTGGGCAACAACATCCACCAGTGGTGTGGTTCCAACAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meshach Asare-Werehene et al.
Oncogene, 39(7), 1600-1616 (2019-11-09)
Ovarian cancer (OVCA) is the most lethal gynecological cancer, due predominantly to late presentation, high recurrence rate and common chemoresistance development. The expression of the actin-associated protein cytosolic gelsolin (GSN) regulates the gynecological cancer cell fate resulting in dysregulation in
Meshach Asare-Werehene et al.
Cancer research, 80(18), 3959-3971 (2020-07-10)
Although initial treatment of ovarian cancer is successful, tumors typically relapse and become resistant to treatment. Because of poor infiltration of effector T cells, patients are mostly unresponsive to immunotherapy. Plasma gelsolin (pGSN) is transported by exosomes (small extracellular vesicle
Zhi-Yuan Chen et al.
Journal of biomedical science, 22, 90-90 (2015-10-21)
Increasing evidence suggests that transforming growth factor-beta 1 (TGF-β1) triggers epithelial to mesenchymal transition (EMT) and facilitates breast cancer stem cell differentiation. Gelsolin (GSN) is a ubiquitous actin filament-severing protein. However, the relationship between the expression level of GSN and
Dylan Z Kelley et al.
Translational research : the journal of laboratory and clinical medicine, 202, 109-119 (2018-08-18)
We have recently performed the characterization of alternative splicing events (ASEs) in head and neck squamous cell carcinoma, which allows dysregulation of protein expression common for cancer cells. Such analysis demonstrated a high ASE prevalence among tumor samples, including tumor-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service