Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU065091

Sigma-Aldrich

MISSION® esiRNA

targeting human LAMP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACCACTGTGCCATCTCCTACTACAACACCTACTCCAAAGGAAAAACCAGAAGCTGGAACCTATTCAGTTAATAATGGCAATGATACTTGTCTGCTGGCTACCATGGGGCTGCAGCTGAACATCACTCAGGATAAGGTTGCTTCAGTTATTAACATCAACCCCAATACAACTCACTCCACAGGCAGCTGCCGTTCTCACACTGCTCTACTTAGACTCAATAGCAGCACCATTAAGTATCTAGACTTTGTCTTTGCTGTGAAAAATGAAAACCGATTTTATCTGAAGGAAGTGAACATCAGCATGTATTTGGTTAATGGCTCCGTTTTCAGCATTGCAAATAACAATCTCAGCTACTGGGATGCCCCCCTGGGAAGTTCTTATATGTGCAACAAAGAGCAGACTGTTTCAGTGTCTGGAGCATTTCAGATAAATACCTTTGATCTAAGGGTTCAGCCTTTCAATGTGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne-Sophie Bach et al.
Oncotarget, 6(29), 28084-28103 (2015-07-18)
The lysosomal protease cathepsin D (Cath-D) is overproduced in breast cancer cells (BCC) and supports tumor growth and metastasis formation. Here, we describe the mechanism whereby Cath-D is accumulated in the nucleus of ERα-positive (ER+) BCC. We identified TRPS1 (tricho-rhino-phalangeal-syndrome
Vikash Singh et al.
The Journal of biological chemistry, 292(5), 1847-1864 (2016-12-10)
Salmonella enterica are invasive intracellular pathogens that replicate within a membrane-bound compartment inside infected host cells known as the Salmonella-containing vacuole. How Salmonella obtains nutrients for growth within this intracellular niche despite the apparent isolation is currently not known. Recent
Anders P Mutvei et al.
Nature communications, 11(1), 1416-1416 (2020-03-19)
The kinase mTOR complex 1 (mTORC1) promotes cellular growth and is frequently dysregulated in cancers. In response to nutrients, mTORC1 is activated on lysosomes by Rag and Rheb guanosine triphosphatases (GTPases) and drives biosynthetic processes. How limitations in nutrients suppress
Chuan-Ying Xu et al.
Frontiers in aging neuroscience, 9, 308-308 (2017-10-13)
α-Synuclein misfolding and aggregation play an important role in the pathogenesis of Parkinson's disease (PD). Loss of function and mutation of the PARK7/DJ-1 gene cause early-onset familial PD. DJ-1 can inhibit α-synuclein aggregation, and may function at an early step

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service