Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU046211

Sigma-Aldrich

MISSION® esiRNA

targeting human KIF2C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTCGGGCTACTTTGGAATGTCATCCACTTACTATGACTGATCCTATCGAAGAGCACAGAATATGTGTCTGTGTTAGGAAACGCCCACTGAATAAGCAAGAATTGGCCAAGAAAGAAATTGATGTGATTTCCATTCCTAGCAAGTGTCTCCTCTTGGTACATGAACCCAAGTTGAAAGTGGACTTAACAAAGTATCTGGAGAACCAAGCATTCTGCTTTGACTTTGCATTTGATGAAACAGCTTCGAATGAAGTTGTCTACAGGTTCACAGCAAGGCCACTGGTACAGACAATCTTTGAAGGTGGAAAAGCAACTTGTTTTGCATATGGCCAGACAGGAAGTGGCAAGACACATACTATGGGCGGAGACCTCTCTGGGAAAGCCCAGAATGCATCCAAAGGGAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haibo Wang et al.
Oncotarget, 8(30), 48671-48687 (2017-04-19)
Defects in resolving kinetochore-microtubule attachment mistakes during mitosis is linked to chromosome instability associated with carcinogenesis as well as resistance to cancer therapy. Here we report for the first time that tumor suppressor p53-binding protein 1 (53BP1) is phosphorylated at
Ashley Mooneyham et al.
Molecular cancer research : MCR, 17(2), 370-383 (2018-10-17)
UNC-45A, a highly conserved member of the UCS (UNC45A/CRO1/SHE4P) protein family of cochaperones, plays an important role in regulating cytoskeletal-associated functions in invertebrates and mammalian cells, including cytokinesis, exocytosis, cell motility, and neuronal development. Here, for the first time, UNC-45A
Chenyu Li et al.
Scientific reports, 6, 18773-18773 (2016-01-07)
Nucleolar and spindle-associated protein (NuSAP) is a microtubule-associated protein that functions as a microtubule stabiliser. Depletion of NuSAP leads to severe mitotic defects, however the mechanism by which NuSAP regulates mitosis remains elusive. In this study, we identify the microtubule
Rui-Chao Chai et al.
Carcinogenesis, 40(10), 1229-1239 (2019-06-04)
1p/19q codeletion, which leads to the abnormal expression of 1p19q genes in oligodendroglioma, is associated with chemosensitivity and favorable prognosis. Here, we aimed to explore the clinical implications of 1p19q gene expression in 1p/19q non-codel gliomas. We analyzed expression of
Hengyi Shao et al.
Scientific reports, 5, 12204-12204 (2015-07-25)
Chromosome segregation in mitosis is orchestrated by the dynamic interactions between the kinetochore and spindle microtubules. The microtubule depolymerase mitotic centromere-associated kinesin (MCAK) is a key regulator for an accurate kinetochore-microtubule attachment. However, the regulatory mechanism underlying precise MCAK depolymerase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service