Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU038051

Sigma-Aldrich

MISSION® esiRNA

targeting human IL23A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGAGGGAACCAAACCAGAGACGCGCTGAACAGAGAGAATCAGGCTCAAAGCAAGTGGAAGTGGGCAGAGATTCCACCAGGACTGGTGCAAGGCGCAGAGCCAGCCAGATTTGAGAAGAAGGCAAAAAGATGCTGGGGAGCAGAGCTGTAATGCTGCTGTTGCTGCTGCCCTGGACAGCTCAGGGCAGAGCTGTGCCTGGGGGCAGCAGCCCTGCCTGGACTCAGTGCCAGCAGCTTTCACAGAAGCTCTGCACACTGGCCTGGAGTGCACATCCACTAGTGGGACACATGGATCTAAGAGAAGAGGGAGATGAAGAGACTACAAATGATGTTCCCCATATCCAGTGTGGAGATGGCTGTGACCCCCAAGGACTCAGGGACAACAGTCAGTTCTGCTTGCAAAGGATCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Jung Kim et al.
Cancer letters, 500, 147-160 (2020-12-06)
Persistent activation of STAT3 and Nrf2 is considered to stimulate the aggressive behavior of basal-like breast cancer (BLBC). However, the precise mechanism underlying sustained overactivation of these transcription factors and their roles in breast cancer progression remain elusive. Analysis of
Tianxiang Wei et al.
Biosensors & bioelectronics, 94, 56-62 (2017-03-05)
MicroRNAs (miRNAs) play important roles in gene regulation and cancer development. Nowadays, it is still a challenge to detect low-abundance miRNAs. Here, we present a magnetic fluorescent miRNA sensing system for the rapid and sensitive detection of miRNAs from cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service