Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU036651

Sigma-Aldrich

MISSION® esiRNA

targeting human STAT5B

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGACGGTGTGATGGAAGTGTTAAAAAAACATCTCAAGCCTCATTGGAATGATGGGGCCATTTTGGGGTTTGTAAACAAGCAACAGGCCCATGACCTACTCATTAACAAGCCAGATGGGACCTTCCTCCTGAGATTCAGTGACTCAGAAATTGGCGGCATCACCATTGCTTGGAAGTTTGATTCTCAGGAAAGAATGTTTTGGAATCTGATGCCTTTTACCACCAGAGACTTCTCCATTCGGTCCCTAGCCGACCGCTTGGGAGACTTGAATTACCTTATCTACGTGTTTCCTGATCGGCCAAAAGATGAAGTATACTCCAAATACTACACACCAGTTCCCTGCGAGTCTGCTACTGCTAAAGCTGTTGATGGATACGTGAAGCCACAGATCAAGCAAGTGGTCCCTGAGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabrielle Sueur et al.
Scientific reports, 10(1), 1906-1906 (2020-02-07)
We recently identified the CDC25A phosphatase as a key actor in proliferation and differentiation in acute myeloid leukemia expressing the FLT3-ITD mutation. In this paper we demonstrate that CDC25A level is controlled by a complex STAT5/miR-16 transcription and translation pathway
Dharmalingam Subramaniam et al.
Cell death & disease, 11(2), 149-149 (2020-02-26)
Osteosarcoma (OS) is the most common primary bone tumor that primarily affects children and adolescents. Studies suggested that dysregulation JAK/STAT signaling promotes the development of OS. Cells treated with pimozide, a STAT5 inhibitor suppressed proliferation and colony formation and induced
Sarah Bertoli et al.
Oncotarget, 6(35), 38061-38078 (2015-10-31)
We investigated cell cycle regulation in acute myeloid leukemia cells expressing the FLT3-ITD mutated tyrosine kinase receptor, an underexplored field in this disease. Upon FLT3 inhibition, CDC25A mRNA and protein were rapidly down-regulated, while levels of other cell cycle proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service