Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU035471

Sigma-Aldrich

MISSION® esiRNA

targeting human PNPLA2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$303.00
50 μG
$571.00

$303.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$303.00
50 μG
$571.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$303.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGTGCCAAGTTCATTGAGGTATCTAAAGAGGCCCGGAAGCGGTTCCTGGGCCCCCTGCACCCCTCCTTCAACCTGGTAAAGATCATCCGCAGTTTCCTGCTGAAGGTCCTGCCTGCTGATAGCCATGAGCATGCCAGTGGGCGCCTGGGCATCTCCCTGACCCGCGTGTCAGACGGCGAGAATGTCATTATATCCCACTTCAACTCCAAGGACGAGCTCATCCAGGCCAATGTCTGCAGCGGTTTCATCCCCGTGTACTGTGGGCTCATCCCTCCCTCCCTCCAGGGGGTGCGCTACGTGGATGGTGGCATTTCAGACAACCTGCCACTCTATGAGCTTAAGAACACCATCACAGTGTCCCCCTTCTCGGGCGAGAGTGACATCTGTCCGCAGGACAGCTCCACCAACATC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosuke Kanno et al.
Arthritis research & therapy, 19(1), 22-22 (2017-02-06)
Systemic sclerosis (SSc) is a connective tissues disease of unknown origin characterized by vascular damage and extensive fibrosis. Recently, we demonstrated that α2-antiplasmin (α2AP) is associated with the development of fibrosis in SSc. We herein investigate the roles of α2AP
Monika Riederer et al.
Archives of physiology and biochemistry, 123(4), 249-253 (2017-04-04)
Vascular endothelial cells represent an important source of arachidonic acid (AA)-derived mediators involved in the generation of anti- or proatherogenic environments. Evidence emerged (in mast cells), that in addition to phospholipases, neutral lipid hydrolases as adipose triglyceride lipase (ATGL) also
Susanne Bürger et al.
International journal of molecular sciences, 22(1) (2021-01-06)
The demise of retinal ganglion cells (RGCs) is characteristic of diseases of the retina such as glaucoma and diabetic or ischemic retinopathies. Pigment epithelium-derived factor (PEDF) is a multifunctional secreted protein that mediates neuroprotection and inhibition of angiogenesis in the
Tian Ma et al.
International journal of oncology, 42(5), 1743-1753 (2013-04-03)
The G0/G1 switch gene 2 (G0S2) is rapidly induced by all-trans-retinoic acid (RA)-treatment of acute promyelocytic leukemia (APL) and other cells. G0S2 regulates lipolysis via inhibition of adipose triglyceride lipase (ATGL). This study found that retinoic acid receptor (RAR), but
Ping Xie et al.
Endocrinology, 156(5), 1648-1658 (2015-03-10)
Intramyocellular accumulation of lipids is often associated with insulin resistance. Deficiency of comparative gene identification-58 (CGI-58) causes cytosolic deposition of triglyceride (TG)-rich lipid droplets in most cell types, including muscle due to defective TG hydrolysis. It was unclear, however, whether

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service