Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU035401

Sigma-Aldrich

MISSION® esiRNA

targeting human CXCL16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGCTTACTCGGGGATTGTGGCCCACCAGAAGCATTTACTTCCTACCAGCCCCCCAATTTCTCAGGCCTCAGAGGGGGCATCTTCAGATATCCACACCCCTGCCCAGATGCTCCTGTCCACCTTGCAGTCCACTCAGCGCCCCACCCTCCCAGTAGGATCACTGTCCTCGGACAAAGAGCTCACTCGTCCCAATGAAACCACCATTCACACTGCGGGCCACAGTCTGGCAGCTGGGCCTGAGGCTGGGGAGAACCAGAAGCAGCCGGAAAAAAATGCTGGTCCCACAGCCAGGACATCAGCCACAGTGCCAGTCCTGTGCCTCCTGGCCATCATCTTCATCCTCACCGCAGCCCTTTCCTATGTGCTGTGCAAGAGGAGGAGGGGGCAGTCACCGCAGTCCTCTCCAGATCTGCCGGTTCATTATATACCTGTGGCACCTGACTCTAATACCTGAGCCAAGAATGGAAGCTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenzhen Ma et al.
Respiratory research, 22(1), 42-42 (2021-02-08)
Alveolar epithelial cells play an essential role in the initiation and progression of pulmonary fibrosis, and the occurrence of epithelial-mesenchymal transition (EMT) may be the early events of pulmonary fibrosis. Recent studies have shown chemokines are involved in the complex
Kun Ling Ma et al.
American journal of translational research, 10(6), 1802-1816 (2018-07-19)
Non-alcoholic fatty liver disease (NAFLD), characterised by early lipid accumulation and subsequent inflammation in the liver, is becoming a worldwide challenge due to its increasing prevalence in developing and developed countries. This study aimed to investigate the role of CXC
Audrey Dieudonné et al.
PloS one, 7(8), e41952-e41952 (2012-08-11)
Scavenger receptors and Toll-like receptors (TLRs) cooperate in response to danger signals to adjust the host immune response. The TLR3 agonist double stranded (ds)RNA is an efficient activator of innate signalling in bronchial epithelial cells. In this study, we aimed
Ze-Bo Hu et al.
Acta pharmacologica Sinica, 39(6), 1022-1033 (2018-04-06)
Inflammation and lipid disorders play crucial roles in synergistically accelerating the progression of diabetic nephropathy (DN). In this study we investigated how inflammation and lipid disorders caused tubulointerstitial injury in DN in vivo and in vitro. Diabetic db/db mice were
Yijie Zhang et al.
Pathology, research and practice, 216(5), 152913-152913 (2020-03-17)
CXC chemokine ligand 16 (CXCL16) has been reported to exacerbate acute kidney injury induced by ischemia-reperfusion (IR). This study aimed to investigate the probable role of CXCL16 in hepatic IR injury during liver transplantation. The expression patterns of CXCL16 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service