Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU031601

Sigma-Aldrich

MISSION® esiRNA

targeting human GPR19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAGGGCTATTCCTGACCAAGAAGATCTAAAGAGAACGTCTCTGAAATCAAGTCCGGATGAAGAATTAAGAGAAAAAAAGTGAATATGGTTTTTGCTCACAGAATGGATAACAGCAAGCCACATTTGATTATTCCTACACTTCTGGTGCCCCTCCAAAACCGCAGCTGCACTGAAACAGCCACACCTCTGCCAAGCCAATACCTGATGGAATTAAGTGAGGAGCACAGTTGGATGAGCAACCAAACAGACCTTCACTATGTGCTGAAACCCGGGGAAGTGGCCACAGCCAGCATCTTCTTTGGGATTCTGTGGTTGTTTTCTATCTTCGGCAATTCCCTGGTTTGTTTGGTCATCCATAGGAGTAGGAGGACTCAGTCTACCACCAACTACTTTGTGGTCTCCATGGCATGTGCTGACCTTCTCATCAGCGTTGCCAGCACGCCTTTCGTCCTGCTCCAGTTCACCACTGGAAGGTGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren M Stein et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 310(6), R476-R480 (2016-01-08)
Adropin, a recently described peptide hormone produced in the brain and liver, has been reported to have physiologically relevant actions on glucose homeostasis and lipogenesis, and to exert significant effect on endothelial function. We describe a central nervous system action
Dharendra Thapa et al.
Redox biology, 18, 25-32 (2018-06-18)
Mitochondria supply ~90% of the ATP required for contractile function in cardiac cells. While adult cardiomyocytes preferentially utilize fatty acids as a fuel source for oxidative phosphorylation, cardiac mitochondria can switch to other substrates when required. This change is driven

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service