Skip to Content
MilliporeSigma
All Photos(1)

Documents

EHU027981

Sigma-Aldrich

MISSION® esiRNA

targeting human GJA5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTTATGGCCAGAAGCCTGAGGTGCCCAATGGAGTCTCACCAGGTCACCGCCTTCCCCATGGCTATCATAGTGACAAGCGACGTCTTAGTAAGGCCAGCAGCAAGGCAAGGTCAGATGACCTATCAGTGTGACCCTCCTTTATGGGAGGATCAGGACCAGGTGGGAACAAAGGAGGCTCAGAGAAGAAAGACGTGTCCCTTCTGAACTGATGCTTTCTCACTGTCATCACTGCTTGGCTCCTTTGAGCCCCGGGTCTCAATGACGTTGCTCATTAATTCTAGAAACTATAACCAGGGCTCTGGGATAGTAAGAGAGGTGACAACCCACCCAGACTGCAGTTCCCTCCCCACCCTCTACCCAGTATACGAAGCCTTTCAGATTACTCATGAAACAGGGTAGAGGGAAAGAAGGGAAGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacques-Antoine Haefliger et al.
Arteriosclerosis, thrombosis, and vascular biology, 37(11), 2136-2146 (2017-10-07)
Cx40 (Connexin40) forms intercellular channels that coordinate the electric conduction in the heart and the vasomotor tone in large vessels. The protein was shown to regulate tumoral angiogenesis; however, whether Cx40 also contributes to physiological angiogenesis is still unknown. Here
Ping Dai et al.
Scientific reports, 4, 7323-7323 (2014-12-05)
The reprogramming of differentiated cells into induced pluripotent stem cells (iPSCs) can be achieved by ectopic expression of defined transcription factors (Oct3/4, Sox2, Klf4 and c-Myc). However, to date, some iPSCs have been generated using viral vectors; thus, unexpected insertional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service