Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU024851

Sigma-Aldrich

MISSION® esiRNA

targeting human CDC6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGCAAAGGCCAGGATGTATTGTACACGCTATTTGAATGGCCATGGCTAAGCAATTCTCACTTGGTGCTGATTGGTATTGCTAATACCCTGGATCTCACAGATAGAATTCTACCTAGGCTTCAAGCTAGAGAAAAATGTAAGCCACAGCTGTTGAACTTCCCACCTTATACCAGAAATCAGATAGTCACTATTTTGCAAGATCGACTTAATCAGGTATCTAGAGATCAGGTTCTGGACAATGCTGCAGTTCAATTCTGTGCCCGCAAAGTCTCTGCTGTTTCAGGAGATGTTCGCAAAGCACTGGATGTTTGCAGGAGAGCTATTGAAATTGTAGAGTCAGATGTCAAAAGCCAGACTATTCTCAAACCACTGTCTGAATGTAAATCACCTTCTGAGCCTCTGATTCCCAAGAGGGTTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zi-Yun Yi et al.
Journal of cellular physiology, 235(7-8), 5541-5554 (2020-01-28)
Cell division cycle protein, CDC6, is essential for the initiation of DNA replication. CDC6 was recently shown to inhibit the microtubule-organizing activity of the centrosome. Here, we show that CDC6 is localized to the spindle from pro-metaphase I (MI) to
Jingwen Zhang et al.
Oncogene, 37(28), 3763-3777 (2018-04-11)
Previous reports have demonstrated that select cancers depend on BRD4 to regulate oncogenic gene transcriptional programs. Here we describe a novel role for BRD4 in DNA damage response (DDR). BRD4 associates with and regulates the function of pre-replication factor CDC6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service