Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU017981

Sigma-Aldrich

MISSION® esiRNA

targeting human GPNMB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTGCAGAAATGAGGCTGGTTTATCTGCTGATCCGTATGTTTACAACTGGACAGCATGGTCAGAGGACAGTGACGGGGAAAATGGCACCGGCCAAAGCCATCATAACGTCTTCCCTGATGGGAAACCTTTTCCTCACCACCCCGGATGGAGAAGATGGAATTTCATCTACGTCTTCCACACACTTGGTCAGTATTTCCAGAAATTGGGACGATGTTCAGTGAGAGTTTCTGTGAACACAGCCAATGTGACACTTGGGCCTCAACTCATGGAAGTGACTGTCTACAGAAGACATGGACGGGCATATGTTCCCATCGCACAAGTGAAAGATGTGTACGTGGTAACAGATCAGATTCCTGTGTTTGTGACTATGTTCCAGAAGAACGATCGAAATTCATCCGACGAAACCTTCCTCAAAGATCTCCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Jin et al.
Oncology reports, 39(6), 3034-3040 (2018-04-06)
Glycoprotein non‑metastatic melanoma protein B (GPNMB) is a glycoprotein that is highly expressed in various types of cancer, including osteosarcoma. However, its cellular functions and related mechanisms in osteosarcoma remain unclear. In the present study, a higher GPNMB mRNA level was
Feifei Ren et al.
Journal of cellular physiology, 235(3), 2738-2752 (2019-09-10)
Gastric cancer has the fifth highest incidence of disease and is the third leading cause of cancer-associated mortality in the world. The etiology of gastric cancer is complex and needs to be fully elucidated. Thus, it is necessary to explore
Basilio Smuczek et al.
Experimental cell research, 358(2), 323-334 (2017-07-10)
Breast cancer is an important public health problem, and its progression may be related to the extracellular matrix (ECM), which acts as a structural scaffold and instruction source for neoplastic cells. Laminins are ECM proteins regulating tumor biology. The laminin-derived
Kazal Boron Biswas et al.
Scientific reports, 10(1), 4930-4930 (2020-03-20)
GPNMB is involved in multiple cellular functions including cell adhesion, stress protection and stem cell maintenance. In skin, melanocyte-GPNMB is suggested to mediate pigmentation through melanosome formation, but details of keratinocyte-GPNMB have yet to be well understood. We confirmed the
Kotaro Kumagai et al.
PloS one, 10(11), e0143413-e0143413 (2015-11-26)
Glycoprotein nonmetastatic melanoma B (Gpnmb), a transmembrane glycoprotein that is expressed in macrophages, negatively regulates inflammation. We have reported that Gpnmb is strongly expressed in the livers of rats fed a choline-deficient, L-amino acid-defined (CDAA) diet. However, the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service