Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU015441

Sigma-Aldrich

MISSION® esiRNA

targeting human ATF6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCCTAGTCCAAAGCGAAGAGTTGTCTGTGTGATGATAGTATTGGCATTTATAATACTGAACTATGGACCTATGAGCATGTTGGAACAGGATTCCAGGAGAATGAACCCTAGTGTGAGCCCTGCAAATCAAAGGAGGCACCTTCTAGGATTTTCTGCTAAAGAGGCACAGGACACATCAGATGGTATTATCCAGAAAAACAGCTACAGATATGATCATTCTGTTTCAAATGACAAAGCCCTGATGGTGCTAACTGAAGAACCATTGCTTTACATTCCTCCACCTCCTTGTCAGCCCCTAATTAACACAACAGAGTCTCTCAGGTTAAATCATGAACTTCGAGGATGGGTTCATAGACATGAAGTAGAAAGGACCAAGTCAAGAAGAATGACAAATAATCAACAGAAAACCCGTATTCTTCAGGGTGCTCTGGAACAGGGCTCAAATTCTCAGCTGATGGCTGTTCAATACACAGAAACCACTAGTAGTATCAGCAGGAACTCAGGGAGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weilin Xu et al.
Frontiers in neuroscience, 12, 638-638 (2018-10-05)
Neuronal apoptosis is an important factor accounting for the poor outcomes of intracerebral hemorrhage (ICH). This study first showed that inhibition of activating transcription factor 6 (ATF6) could alleviate secondary brain injury through anti-apoptosis after ICH in rats. Melatonin, ATF6
Patricia Freis et al.
Oncotarget, 8(13), 20974-20987 (2017-04-21)
mTOR and Unfolded Protein Response (UPR) are two signaling pathways frequently activated in cancer cells. The mTOR pathway has been shown to be up-regulated in most gastroenteropancreatic neuroendocrine tumors. In contrast, little is known about the UPR status in neoplastic
Rui Zhou et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 51(5), 2397-2420 (2018-12-12)
Lipid droplets (LDs) are dynamic organelles that store neutral lipids during times of energy excess, and an increased accumulation of LDs in the liver is closely linked to hepatic steatosis. Our previous studies suggested that resveratrol (RSV) supplement could improve
A M Merlot et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(9), 2094-2110 (2019-04-15)
The metastasis suppressor, N-myc downstream regulated gene-1 (NDRG1), is a stress response protein that is involved in the inhibition of multiple oncogenic signaling pathways. Initial studies have linked NDRG1 and the endoplasmic reticulum (ER) stress response. Considering this, we extensively
To Sing Fung et al.
Virology, 533, 34-44 (2019-05-15)
Coronavirus infection induces the generation of autophagosomes, and certain host proteins regulating cellular autophagy are hijacked by some coronaviruses to facilitate the formation of double membrane vesicles. However, mechanisms underlying coronavirus-induced autophagy remain largely unknown. In this study, we demonstrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service