Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU015141

Sigma-Aldrich

MISSION® esiRNA

targeting human ATG3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGGGCTACAGGGGAAGAATTGAAAGTGAAGGCATACCTACCAACAGGCAAACAATTTTTGGTAACCAAAAATGTGCCGTGCTATAAGCGGTGCAAACAGATGGAATATTCAGATGAATTGGAAGCTATCATTGAAGAAGATGATGGTGATGGCGGATGGGTAGATACATATCACAACACAGGTATTACAGGAATAACGGAAGCCGTTAAAGAGATCACACTGGAAAATAAGGACAATATAAGGCTTCAAGATTGCTCAGCACTATGTGAAGAGGAAGAAGATGAAGATGAAGGAGAAGCTGCAGATATGGAAGAATATGAAGAGAGTGGATTGTTGGAAACAGATGAGGCTACCCTAGATACAAGGAAAATAGTAGAAGCTTGTAAAGCCAAAACTGATGCTGGCGGTGAAGATGCTATTTTGCAAACCAGAACTTATGACCTTTACATCACTTATGATAAATATTACCAGACTCCACGATTATGGTTGTTTGGCTATGATGAGCAACG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiuli Zhang et al.
Journal of molecular neuroscience : MN, 71(2), 284-292 (2020-07-20)
This study aimed to clarify the regulation role of miR-708 and miR-335-3p in retinal ganglion cell (RGC) autophagy and apoptosis in glaucoma. Chronic glaucoma mice were established by laser photocoagulation. RGCs were isolated and transfected with a series of plasmids
Alexander Agrotis et al.
Autophagy, 15(6), 976-997 (2019-01-22)
Macroautophagy/autophagy is a cellular degradation pathway that delivers cytoplasmic material to lysosomes via double-membrane organelles called autophagosomes. Lipidation of ubiquitin-like LC3/GABARAP proteins on the autophagosome membrane is important for autophagy. The cysteine protease ATG4 executes 2 LC3/GABARAP processing events: priming

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service