Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU013041

Sigma-Aldrich

MISSION® esiRNA

targeting human KDM4A

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$315.00
50 μG
$596.00

$315.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
$315.00
50 μG
$596.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$315.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACTGCAGGCCAGAAAGTCATTAGCAAGCATAAGAACGGGCGCTTCTACCAGTGTGAAGTGGTCAGGCTCACCACCGAGACCTTCTATGAAGTCAACTTTGATGATGGCTCCTTCAGCGACAATCTTTATCCTGAGGACATAGTGAGCCAGGACTGTCTCCAGTTTGGTCCTCCTGCTGAAGGGGAAGTGGTCCAAGTGAGATGGACAGACGGCCAAGTCTATGGAGCCAAGTTTGTGGCCTCCCACCCTATCCAAATGTACCAGGTGGAGTTTGAGGATGGCTCACAACTTGTGGTTAAGAGAGATGATGTATACACACTGGATGAAGAGCTTCCCAAGAGAGTCAAATCTAGACTGTCAGTAGCCTCAGACATGCGCTTCAATGAGATTTTCACAGAGAAAGAGGTTAAGCAAGAAAAGAAACGGCAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Su et al.
BMC cancer, 17(1), 477-477 (2017-07-12)
Jumonji C domain 2A (JMJD2A), as a histone demethylases, plays a vital role in tumorigenesis and progression. But, its functions and underlying mechanisms of JMJD2A in nasopharyngeal carcinoma (NPC) metabolism are remained to be clarified. In this study, we investigated
Antonio Pezone et al.
Nucleic acids research, 48(16), 8943-8958 (2020-07-23)
The epithelial-to-mesenchymal transition (EMT) is a complex transcriptional program induced by transforming growth factor β1 (TGF-β1). Histone lysine-specific demethylase 1 (LSD1) has been recognized as a key mediator of EMT in cancer cells, but the precise mechanism that underlies the
Haiyu Zhang et al.
Archives of biochemistry and biophysics, 684, 108334-108334 (2020-03-17)
Emerging evidence shows that histone modification and its related regulators are involved in the progression and chemoresistance of ovarian cancer (OC) cells. Our present study found that the expression of Jumonji C domain-containing 2A (JMJD2A), while not JMJD2B or JMJD2C
Tadahiko Nakagawa et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 23(3), 426-436 (2019-11-05)
Jumonji domain-containing protein 2A (JMJD2A) of the JMJD2 family of histone lysine demethylases has been implicated in tumorigenesis. However, its expression and role in gastric cancer (GC) drug resistance remain unknown. Here, we investigated the role of JMJD2A in GC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service