Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU010441

Sigma-Aldrich

MISSION® esiRNA

targeting human VDR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAGTTCGTGTGAATGATGGTGGAGGGAGCCATCCTTCCAGGCCCAACTCCAGACACACTCCCAGCTTCTCTGGGGACTCCTCCTCCTCCTGCTCAGATCACTGTATCACCTCTTCAGACATGATGGACTCGTCCAGCTTCTCCAATCTGGATCTGAGTGAAGAAGATTCAGATGACCCTTCTGTGACCCTAGAGCTGTCCCAGCTCTCCATGCTGCCCCACCTGGCTGACCTGGTCAGTTACAGCATCCAAAAGGTCATTGGCTTTGCTAAGATGATACCAGGATTCAGAGACCTCACCTCTGAGGACCAGATCGTACTGCTGAAGTCAAGTGCCATTGAGGTCATCATGTTGCGCTCCAATGAGTCCTTCACCATGGACGACATGTCCTGGACCTGTGGCAACCAAGACTACAAGTACCGCGTCAGTGACGTGACCAAAGCCGGACACAGCCTGGAGCTGATTGAGCCCCTCATCAAGTTCCAGGTGGGACTGAAGAAGCTGAACTTGCATGAGGAGGAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Gao et al.
International journal of molecular medicine, 46(4), 1538-1550 (2020-09-19)
Psoriasis is an immune‑mediated dermatosis characterized by T‑lymphocyte‑mediated epidermal hyperplasia, for which there are currently no effective clinical treatments. 'Psoriasis 1' is a Chinese herbal medicine formulation that has been recently used extensively in China for treating patients with psoriasis. However
Ken-ichiro Tanaka et al.
Biochemical and biophysical research communications, 450(1), 482-487 (2014-06-14)
Vitamin D deficiency and advanced glycation end products (AGEs) are suggested to be involved in the pathogenesis of osteoporosis and sarcopenia. However, the effects of vitamin D and AGEs on myogenesis and the interaction between muscle and bone remains still
Lars Brodowski et al.
PloS one, 9(6), e98527-e98527 (2014-06-03)
Placenta-derived circulating factors contribute to the maternal endothelial dysfunction underlying preeclampsia. Endothelial colony forming cells (ECFC), a sub-population of endothelial progenitor cells (EPCs), are thought to be involved in vasculogenesis and endothelial repair. Low vitamin D concentrations are associated with
Aurélien Mary et al.
Endocrinology, 156(6), 1965-1974 (2015-03-13)
Vascular calcification (VC) is a degenerative disease that contributes to cardiovascular morbidity and mortality. A negative relationship has been demonstrated between VC and calcium sensing receptor (CaSR) expression in the vasculature. Of interest, vitamin D response elements, which allow responsiveness
T P H Nguyen et al.
Journal of molecular medicine (Berlin, Germany), 93(7), 795-805 (2015-02-27)
Fetal growth restriction (FGR) affects up to 5 % of pregnancies worldwide, and trophoblast function plays a significant role on the outcome. An epidemiological study has linked vitamin D deficiency to adverse perinatal outcomes, which include decreased birth weight. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service